Learn More
Invitrogen™ Gateway™ BP Clonase™ Enzyme Mix
Brand: Invitrogen™ 11789013
764.03 EUR valid until 2024-09-28
Use promo code "21626" to get your promotional price.
Alert:
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21626” to get your promotional price
Includes: Proteinase K solution (2μg/μL), 30% PEG 8000/30mM MgCl2 Solution and a positive control vector.
Description
- Proteins catalyze in vitro recombination of PCR products or DNA segments from clones (containing attB sites) and Donor vector (containing attP sites) to generate Entry clones
- BP Clonase II enzyme format is single mix of both enzyme and buffer to ensure enzyme stability and enable convenient 10μL reaction set up with fewer pipetting steps, which both maximize recombination efficiency and minimize pippetting
- Original format of BP Clonase enzyme is stand alone enzyme blend and reaction buffer provided in separate tubes
Cloning, Gateway Cloning, Gateway Recombination
Order Info
Shipping Conditions: Dry ice
Specifications
All Gateway™ BP Clonase™ enzyme kits include proteinase K solution (2 μg/μl), 30% PEG 8000/30 mM MgCl2 Solution and a positive control vector. Store BP Clonase™ enzyme at -80°C; BP Clonase™ II Enzyme mixtures at -20°C. Guaranteed stable for 6 months when properly stored. |
|
BP Clonase | |
20 Reactions | |
Clonase™, Gateway™ |
Dry Ice | |
Reaction Buffer | |
BP Clonase Enzyme Mix |
For Research Use Only. Not for use in diagnostic procedures.