missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Gateway™ BP Clonase™ II Enzyme mix

Catalyzes in vitro recombination of PCR products to generate entry clones

Brand:  Invitrogen™ 11789100

 View more versions of this product

Product Code. 10348582

  • 1552.00 € / 100 reactions

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

Includes: Proteinase K solution (2μg/μL) and a positive control vector
This item is not returnable. View return policy

Description

Description

  • Catalyzes the in vitro recombination of PCR products or subcloning DNA segments from clones (containing attB sites) and a donor vector (containing attP sites) to generate entry clones
  • Contains enzymes and buffer in a single mix to enable convenient ten-microliter reaction set up with fewer pipetting steps
  • Guaranteed stable for 6 months when properly stored at -20°C to -80°C

Order Info

Shipping Conditions: Dry ice

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Gateway™ BP Clonase™ II enzyme mix includes proteinase K solution (2 μg/μl) and a positive control vector. Store at -20°C or -80°C. Guaranteed stable for 6 months when properly stored.
BP Clonase
100 Reactions
Clonase™, Gateway™
Dry Ice
Enzyme Buffer
BP Clonase Enzyme Mix
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.