missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Gateway™ LR Clonase™ Enzyme mix
Enzyme mix that offers high recombination efficiency
Brand: Invitrogen™ 11791019
486.08 EUR valid until 2024-11-30
Use promo code "21651" to get your promotional price.
Includes: Proteinase K solution (2μg/μL) and a positive control vector
Description
Gateway LR Clonase enzyme mix contains proprietary blend of Int (Integrase), IHF (Integration Host Factor) and Xis (Excisionase) enzymes that catalyze the in vitro recombination between an Entry clone (containing a gene of interest flanked by attL sites) and a Destination vector (containing attR sites) to generate your expression clone.
- Specifically optimized for both single and multiple fragment cloning
- Provided in optimized single mix of enzyme and reaction buffer, ensuring enzyme stability and ease of use with few pippetting steps for both single and multiple fragment cloning
Cloning, Gateway Cloning, Gateway Recombination
Order Info
Shipping Conditions: Dry ice
Specifications
All Gateway™ LR Clonase™ enzyme kits include proteinase K solution (2 μg/μl) and a positive control vector. Store LR Clonase™ enzyme at -80°C; LR Clonase™ II or II Plus Enzyme mixtures at -20°C. Guaranteed stable for 6 months when properly stored. | |
LR Clonase | |
20 Reactions | |
Clonase™, Gateway™ |
Dry Ice | |
Reaction Buffer | |
LR Clonase Enzyme Mix |
For Research Use Only. Not for use in diagnostic procedures.