missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Gateway™ LR Clonase™ II Enzyme mix
Contains enzymes and buffer in a single mix to enable convenient ten-microliter reaction set-up with fewer pipetting steps.
374.00 € - 1526.00 €
Specifications
Content And Storage | Gateway™ LR Clonase™ II enzyme mix includes proteinase K solution (2 μg/μL) and a positive control vector. Store at -20°C or -80°C. Guaranteed stable for 6 months when properly stored. |
---|---|
Shipping Condition | Dry Ice |
Enzyme | LR Clonase |
Compatible Buffer | Enzyme Buffer |
Product Type | LR Clonase Enzyme Mix |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10134992
|
Invitrogen™
11791020 |
20 Reactions |
374.00 €
20 reactions |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10033092
|
Invitrogen™
11791100 |
100 Reactions |
1526.00 €
100 reactions |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Gateway™ LR Clonase™ II enzyme mix catalyzes the in vitro recombination between an entry clone (containing a gene of interest flanked by attL sites) and a destination vector (containing attR sites) to generate an expression clone
- It contains enzymes and buffer in a single mix to enable convenient ten-microliter reaction set-up with fewer pipetting steps
- Guarenteed stable for 6 months when properly stored at -20°C or -80°C
Order Info
Shipping Conditions: Dry ice
Specifications
Gateway™ LR Clonase™ II enzyme mix includes proteinase K solution (2 μg/μL) and a positive control vector. Store at -20°C or -80°C. Guaranteed stable for 6 months when properly stored. | |
LR Clonase | |
LR Clonase Enzyme Mix |
Dry Ice | |
Enzyme Buffer | |
Clonase™, Gateway™ |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Gateway LR Clonase II Enzyme mix
Signal Word
- Danger
Hazard Category
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Respiratory sensitiser Category 1
Hazard Statement
- H301-Toxic if swallowed.
- H311-Toxic in contact with skin.
- H331-Toxic if inhaled.
- H314-Causes severe skin burns and eye damage.
- H372-Causes damage to organs.
- H361-Suspected of damaging fertility or the unborn child.
- H351-Suspected of causing cancer.
- H341-Suspected of causing genetic defects.
- H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P362-Take off contaminated clothing.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P501-Dispose of contents/container to .
Supplemental information
- MIXTURE LIST-Contains : Glycerin, 1, 4-Butanediamine, N-(3-aminopropyl)-, Proteinase, tritirachium albumserine
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title