Learn More
Invitrogen™ Countess™ Automated Cell Counter, with 101 Boxes of Slides
Benchtop instrument for easy cell and viability counts in as little as 30 seconds
Brand:  Invitrogen™ C10311
 View more versions of this product
Product Code. 10497698
  • 17390.00 € / Each
Explore more special offers
Includes:
  • One Countess Automated Cell Counter, 101 boxes of 50 slides each (1050 total slides), 2 x 100mL bottles of Trypan blue stain, Power cord with 4 adaptors and Quick Reference Guide (C10311)
  • One Countess Automated Cell Counter, one box of 50 slides (100 total slides), 2 x 1mL vials of Trypan blue stain, power cord with 4 adaptors, and Quick Reference Guide (C10227)
  • One Countess Automated Cell Counter, 11 boxes of 50 slides each (550 total slides), 22 x 1mL vials of Trypan blue stain, Power cord with 4 adaptor and Quick Reference Guide (C10310)
Description
Description
  • For verifying Number of Cells, Live Cells/mL, Dead Cells/mL, Cell Size
  • Compatible with Countess cell counting chamber slides
  • Dimensions: D x W x H: 20 x 27 x 19cm (7.8 x 10.6 x 7.4 in.)
Cell Counting, Viability and Cryopreservation
TRUSTED_SUSTAINABILITY
Specifications
Specifications
20 x 27 x 19cm
50/60 Hz
One Countess Automated Cell Counter, 101 boxes of 50 slides each (1050 total slides), 2 x 100mL bottles of Trypan blue stain, Power cord with 4 adaptors and Quick Reference Guide
100-240 VAC
Automated Cell Counter, 101 boxes of slides
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Invitrogen™ Countess™ Automated Cell Counter, with 101 Boxes of Slides > Automated Cell Counter with 101 boxes of slides