missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ dCTP (100 mM)

Invitrogen dCTP is prepared to a final concentration of 100 mM in highly purified water. 100 mM dCTP is suitable for use in polymerase chain reaction (PCR), sequencing, fill-in reactions, nick translation, cDNA synthesis, and TdT-tailing reactions.

Brand:  Invitrogen™ 10217016

132.65 EUR valid until 2025-06-30
Use promo code "24074" to get your promotional price.


Product Code. 10267632

  • 176.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

Includes: Supplied at a concentration of 100mM in highly purified water (pH 7.5).
This item is not returnable. View return policy

Description
Show More
Specifications

Specifications

100 mM dCTP Solution in highly purified water (250 μL)

Store at –20°C.
Unlabeled
Approved for shipment on Wet or Dry Ice
100 mM
Liquid
250 μL
Standard PCR, High-fidelity PCR, RT-PCR, Real Time PCR (qPCR)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Manuals

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ dCTP (100 mM) > 250uL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.