missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DNase I, Amplification Grade

Purified and tested for nondetectable levels of RNase contamination

Brand:  Invitrogen™ 18068015

Product Code. 10694233

  • 239.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

Includes: DNase I, Amp Grade (100U), 10X DNase I Reaction Buffer, 25mM EDTA (pH 8.0)
This item is not returnable. View return policy

Description

Description

DNase I, Amplification Grade, digests single- and double-stranded DNA to oligodexyribonuleotides containing a 5' phosphate. DNase I, Amplification Grade, is suitable for eliminating DNA during critical RNA purification procedures such as those prior to RNA-PCR amplification. Absence of RNase is tested by performing a ribonuclease assay with RNA ladder.

  • Specific activity is >10,000 units/mg
  • Source: Purified from bovine pancreas
  • Performance and quality testing: Ribonuclease assay with RNA ladder and ability to digest single-stranded and double-stranded DNA to oligonucleotides are determined
  • Unit definition: One unit increases absorbance of high molecular weight DNA solution at rate of 0.001 A260 units/min./mL of reaction mixture at 25°C
  • Unit reaction conditions: 0.1M sodium acetate (pH 5.0), 5mM MgCl2, 50 μg/mL calf thymus DNA, and enzyme in 1mL for 10 min. at 25°C

Removing DNA from RNA and protein preparations, PCR & Real-Time PCR, Real Time PCR (qPCR), Reverse Transcription

Order Info

Shipping Conditions: Approved for shipment on wet or dry ice

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Contents:
1 vial of DNase I, Amp Grade (100 U)
1 vial of 10X DNase I Reaction Buffer (1000 μL)
1 vial of 25 mM EDTA (pH 8.0) (200 μL)

Store at -20°C in a non-frost-free freezer.
DNase
100 U
Approved for shipment on Wet or Dry Ice
Reaction Buffer
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.