missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DNase I
Purified from bovine pancreas
Brand: Invitrogen™ 18047019
236.21 EUR valid until 2025-03-28
Use promo code "24107" to get your promotional price.
Includes: DNase I is supplied as 1 vial containing 20,000 units at a concentration of 50-375U/μL.
Description
- DNase I (Deoxyribonuclease I) digests single- and double-stranded DNA to oligodeoxyribonucleotides containing a 5' phosphate
- Ribonuclease has been reduced to nondetectable levels
- Specific activity of DNase I is typically in range of 10,000–25,000 units/mg
For removing DNA from RNA preparations, use Amplification Grade DNase I (Mfr. No. 18068-015).
Removing DNA from protein preparations, Nick translating DNA, Generating random fragments for dideoxy sequencing, PCR & Real-Time PCR, Reverse transcription
Order Info
Shipping Conditions: Wet ice
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
50 to 375 U/μL | |
Wet Ice | |
20,000 U |
DNase I is supplied as 1 vial containing 20,000 units at a concentration of 50-375 U/μl. Store in freezer (-5°C to -30°C). | |
DNase | |
DNase I |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?Share a Content Correction