missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DNAzol™ Reagent, for isolation of genomic DNA from solid and liquid samples

DNAzol Reagent is a complete and ready-to-use organic reagent from animal, plant, yeast, and bacterial origin.

Brand:  Invitrogen™ 10503027

Product Code. 11558626

  • 411.00 € / 100mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

Includes: 1 bottle (100mL)
This item is not returnable. View return policy

Description

Description

DNAzol Reagent is a complete and ready-to-use organic reagent from animal, plant, yeast, and bacterial origin. The DNAzol Reagent procedure can be completed in typically 10–30 minutes with DNA recoveries of 70–100%.

  • Versatile: Can use with broad spectrum of sample materials
  • Rapid isolation and high recovery of genomic DNA
  • Efficient isolation of genomic DNA from a variety of sample types
  • Rapid isolation and high recovery of genomic DNA
  • Isolated DNA can be used for a number of downstream applications

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY
Specifications

Specifications

100 mL DNAzol Reagent; room temperature

Bacteria, Cells, Plant, Tissue, Yeast
Genomic DNA
Not High-throughput Compatible (Manual)
Room Temperature
Bacteria: ≤107 cells or 0.1 mL
Cells: ≤107
Plant: ≤50 mg
Tissue: ≤50 mg
Yeast: ≤107 cells or 0.1 mL
Organic Extraction
Varies
PCR, southern blotting, sequencing, nucleic acid labeling, hybridization
100 mL
30 min.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.