missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Exosome-Human CD9 Flow Detection Reagent (from cell culture)

Cell culture media must first be enriched for exosomes using Total Exosome Isolation Reagent (from cell culture media) or traditional ultracentrifugation.

Brand:  Invitrogen™ 10620D

Product Code. 15480544

  • 248.00 € / 2mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Cell culture media must first be enriched for exosomes using Total Exosome Isolation Reagent (from cell culture media) or traditional ultracentrifugation. Exosome-Human CD9 Flow Detection Reagent (from cell culture) allows easy visualization of exosomes by flow cytometry using this magnetic separation technology. Free exosomes alone are too small to be detected by most flow cytometry instruments. The 2.7 μm Dynabeads magnetic beads allow for a clear and defined flow analysis.

  • Minimal hands-on time for specific exosome capture and analysis
  • Clear and defined flow analysis in less than 1 hour hands-on time
  • Study expression of specific exosome subset markers
  • Protocol is easily scalable
  • Recent studies suggest that exosome/ECV population consists of several subsets, possibly with different functions
  • Using immuno-affinity based Dynabeads magnetic separation technology, these subsets can be selectively captured and studied using flow cytometry and/or electron microscopy
  • Dynabeads technology avoids loss of material due to handling and centrifugation typically experienced with other isolation and analysis methods

Mix thoroughly to ensure that magnetic beads do not settle in the sample tube. A mixer which provides both tilting and rotation for optimal mixing is recommended.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Store in refrigerator (2–8°C).
Dynabeads™
Cell Cultures
Flow Detection Reagent
2 mL
2.7 μm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Manuals

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2759045 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number10620D
Lot Number2812140 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number10620D
Lot Number2812142 Certificate TypeCertificate of Analysis Date28/01/2025 Catalog Number10620D
Lot Number2882219 Certificate TypeCertificate of Analysis Date28/01/2025 Catalog Number10620D
Lot Number2906523 Certificate TypeCertificate of Analysis Date26/01/2025 Catalog Number10620D
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ Exosome-Human CD9 Flow Detection Reagent (from cell culture) > 2mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.