missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Dynabeads™ M-270 Carboxylic Acid
These are hydrophilic Dynabeads™ (2.8 μm) with carboxylic acid groups. These surface groups allow covalent amide bond formation to proteins/peptides via primary amino- or sulphydryl groups.
Brand: Invitrogen™ 14305D
This item is not returnable.
View return policy
Description
These surface groups allow covalent amide bond formation to proteins/peptides via primary amino- or sulphydryl groups. For use with any sample type.
- Immediate and gentle coupling at low temperature and neutral to low pH
- Activation through carbodiimide is required
- Alternative reactive groups can be introduced by using commercially available cross-linkers
- Used for isolation of small components such as proteins and peptides
- Immobilization of labile proteins/peptides and N-terminal coupling of peptides
- Low non-specific binding of nucleic acids
- High throughput compatible
Order Info
Shipping Condition: Room Temperature
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Store in refrigerator (2–8°C). | |
5 μL | |
High-throughput Compatible | |
Research Grade | |
Any Sample Type | |
Carboxylic Acid |
Carboxylic Acid Magnetic Bead | |
Beads in suspension | |
DYNAL™, Dynabeads™ | |
2 mL | |
Room Temperature | |
2.8 μm |
For Research Use Only. Not for use in diagnostic procedures.