Learn More
Invitrogen™ Dynabeads™ Goat Anti-Mouse IgG
Use with any mouse IgG for depletion, or for positive isolation of cells for molecular purposes.
Brand: Invitrogen™ 11033
Includes: 5mL Dynabeads coated with goat polyclonal against all mouse IgG subclasses.
Description
Use with any mouse IgG for depletion, or for positive isolation of cells for molecular purposes. Any cell can be isolated from any samples from all species, except mouse and used in further molecular analysis.
• Deplete any target cell from all species, except mouse
• Isolate cells directly from whole blood for molecular applications
• Use a cocktail of mouse IgG's to deplete unwanted labeled cells and obtain untouched, bead and antibody-free cells.
This Product Contains:
5 mL Dynabeads™ coated with goat polyclonal against all mouse IgG subclasses.
Starting samples:
Whole blood, cord blood, MNC, buffy coat, mouse spleen/ lymph nodes, tissue digests.
Order Info
Shipping Condition: Room Temperature
Specifications
Whole Antibody | |
Polyclonal | |
Antibody Coated Bead | |
Goat | |
Processes ∼2 x 109 cells total | |
DYNAL™, Dynabeads™ | |
5 mL | |
PBMC, Tissue Digests, Blood | |
1 x 107 PBMCs per isolation |
All cells from all species, except mouse | |
This Product Contains: 5 mL Dynabeads™ coated with goat polyclonal against all mouse IgG subclasses. Store at 2°C to 8°C. |
|
Dynabeads™ | |
Depletion, negative isolation or positive isolation | |
>95% | |
Research Grade | |
All species, except mouse | |
Room Temperature | |
All species, except mouse |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.