missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Dynabeads™ M-270 Epoxy

Dynabeads™ M-270 Epoxy beads are 2.8 μm superparamagnetic beads containing surface epoxy groups.

Brand:  Invitrogen™ 14301

426.70 EUR valid until 2024-12-31
Use promo code "21651" to get your promotional price.


 View more versions of this product

Product Code. 10512625

  • 502.00 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket

Includes: 60mg Dynabeads M-270 Epoxy, approx. 6.7 x 107 beads/mg, supplied as a freeze-dried powder.
This item is not returnable. View return policy

Description

Description

Dynabeads™ M-270 Epoxy beads are 2.8 μm superparamagnetic beads containing surface epoxy groups. The beads covalently bind primary amino and sulfhydryl groups in proteins and peptides, making them ideal for coupling antibodies, peptides, intact proteins, and functional enzymes. These hydrophylic, neutral pH beads exhibit extremely low nonspecific binding of proteins and dyes, which reduces the need for blocking agents. Also, Dynabeads™ M-270 Epoxy Beads do not contain Tween™ detergent, and so are ideal for use in mass spectrometry (MS) analysis.

• Ideal for immunoprecipitation (IP) of proteins and protein complexes
• Gentle, yet rapid magnetic separation and short incubation times allow identification of transient and labile complexes
• Covalent antibody-coupling to the Dynabeads™ avoids co-elution of the antibody with the target protein

Adaptable for various applications
Fast surface binding kinetics and rapid magnetic separation characteristics make Dynabeads™ M-270 Epoxy beads the ideal choice for applications such as immunoprecipitation/purification of proteins and protein complexes, coupling functional enzymes to the bead surface for downstream assays, and identifying protein binding partners. Captured proteins and protein complexes are easily separated, washed, and eluted using a DynaMag™ magnet, and the magnetic separation properties of the Dynabeads™ .

Coupling procedure
Covalent coupling is performed overnight by incubating the desired ligand (commonly antibodies, peptides, or proteins) with Dynabeads™ M-270 Epoxy beads. The binding capacity varies depending on the ligand used, but is typically 5–10 μg IgG per milligram of beads. Coupling occurs at neutral pH and high salt concentration. The coupling reaction can be performed over a broad range of temperatures to match the stability characteristics of your ligand.

Upon completion of the ligand coupling step, the reactive moieties on the Dynabeads™ M-270 Epoxy beads are deactivated to prevent further coupling. Once the coupling reaction is completed, these nonporous, hydrophilic, pH neutral beads exhibit extremely low nonspecific binding.

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Contains 60 mg Dynabeads™ M-270 Epoxy, ∼6.7 x 107 beads/mg, supplied as a freeze-dried powder.

Store at 2°C to 8°C.
5 μL
Any Sample Type
DYNAL™, Dynabeads™
Epoxy Bead
Research Grade
2.8 μm
Room Temperature
Epoxy
60 mg
Freeze Dried
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.