Learn More
Invitrogen™ Dynabeads™ M-270 Streptavidin
Dynabeads M-270 Streptavidin beads are 2.8-μm monosized, monodispersed, hydrophilic, superparamagnetic beads covalently coupled to streptavidin.
Brand: Invitrogen™ 65305
478.69 EUR valid until 2024-09-28
Use promo code "21626" to get your promotional price.
Alert:
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21626” to get your promotional price
Includes: Dynabeads are supplied in PBS, pH 7.4 with 0.01% Tween™-20 and 0.02% NaN3 added as preservatives
Description
Dynabeads M-270 Streptavidin beads are 2.8-μm monosized, monodispersed, hydrophilic, superparamagnetic beads covalently coupled to streptavidin. The product is without blocking and intended for use in the isolation and handling of biotinylated nucleic acids, antibodies, and other biotinylated ligands. The excellent dispersion abilities and the absence of magnetic remanence make the Dynabeads streptavidin products ideal for both manual and automated protocols, including microfluidic system protocols.
Benefits of Dynabeads M-270 Streptavidin beads include:
• Performance—high sensitivity and reproducibility secure consistent results
• Efficiency—fast binding kinetics and high binding capacity
• Flexibility—flexible protocols with gentle and efficient liquid-phase reaction kinetics
• Automation ready—easy automation for high-throughput needs on the Kingfisher Purification System
Consistency and reproducibility
The monolayer of streptavidin leaves most of the biotin-binding sites available for the binding of free biotin and biotinylated ligands. The specific and defined surface allows for efficient capture, separation, and downstream handling. In addition, the low size distribution (<5%) of the monosized beads and the streptavidin monolayer ensure negligible leakage, providing batch consistency and reproducibility of results.
High binding capacity
The size of the ligand and the biotinylation procedure will affect the binding capacity. The capacity also depends on steric availability and charge interaction between bead and ligand and between ligands. With two or three biotin binding sites available for each streptavidin molecule on the surface of the bead after immobilization, one mg of Dynabeads M-270 Streptavidin typically binds:
• ∼ >950 pmol free biotin
• ∼10 μg biotinylated IgG
• ∼200 pmol biotinylated peptides
• ∼10 μg biotinylated ds-DNA
• ∼200 pmol biotinylated ss-oligonucleotides
Applications
Thermo Fisher streptavidin-coupled Dynabeads magnetic beads have been used and cited for a wide variety of applications. Although the beads can be used to isolate any biotinylated targets, these hydrophilic beads are particularly well suited for the preparation of single-stranded DNA, use in protocols that require GTC lysis or high salt concentrations and immunoassays.
Automation ready with the KingFisher Purification System
The 2.8-μm magnetic beads are ideal for high-throughput enrichment. This process can be automated using any of the KingFisher Sample Purification Systems or liquid handlers.
Commercial supply
Our manufacturing sites are ISO 13485-certified. Bead characteristics and manufacturing conditions help ensure consistency and batch reproducibility making them an ideal choice for commercial supply. If you are in the process of customizing Dynabeads M-270 Streptavidin beads in a commercial product or service or need a larger bulk volume of this product, please contact us at oemdynal@lifetech.com or see our Dynabeads OEM page.
Order Info
Shipping Condition: Room Temperature
Specifications
ISO9001 and ISO13485 | |
Dynabeads M-270 Streptavidin is supplied in PBS, pH 7.4 with 0.02% NaN3. Store at 2°C to 8°C. |
|
Capture of Biotinylated Targets | |
Streptavidin Magnetic Beads | |
KingFisher™ Sample Purification System, DynaMag™ magnets | |
Magnetic Beads | |
0.02% NaN3 | |
Biotinylated nucleic acids, antibodies, or other biotinylated ligands and target | |
Ambient Temperature | |
Monosized 2.8 μm (CV <5%) | |
2.8 μm |
10 mg/mL | |
Recombinant streptavidin covalently coupled to Dynabeads | |
Magnetic Polystyrene | |
Beads in suspension | |
High-throughput Compatible | |
Streptavidin | |
Dynabeads™ | |
Any Sample Type | |
Carboxylic Acid, Hydrophilic | |
2 mL | |
For Research Use Only |
For Research Use Only. Not for use in diagnostic procedures.