missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Dynabeads™ M-280 Streptavidin

Dynabeads M-280 Streptavidin beads are 2.8-μm monosized, monodispersed, superparamagnetic, and hydrophobic beads covalently coupled to streptavidin.

Brand:  Invitrogen™ 11205D

606.90 EUR valid until 2024-12-20
Use promo code "21651" to get your promotional price.


 View more versions of this product

Product Code. 10292593

  • 714.00 € / 2mL
In Stock
to see stock.
Edge
Explore more special offers
Add to basket

Includes: Dynabeads (2.8μm) with covalently coupled recombinant streptavidin. Supplied in PBS, pH 7.4 with 0.1% BSA and 0.02% sodium azide added as a preservative
This item is not returnable. View return policy

Description

Description

Dynabeads M-280 Streptavidin beads are 2.8-μm monosized, monodispersed, superparamagnetic, and hydrophobic beads covalently coupled to streptavidin. They are blocked with BSA and intended for use in the isolation and handling of biotinylated nucleic acids, antibodies, and other biotinylated ligands. The excellent dispersion abilities and the absence of magnetic remanence make the Dynabeads streptavidin products ideal for both manual and automated protocols, including microfluidic system protocols.

Benefits of Dynabeads M-280 Streptavidin beads include:
Performance—high sensitivity and reproducibility secure consistent results
Efficiency—fast binding kinetics and high binding capacity
Flexibility—flexible protocols with gentle and efficient liquid-phase reaction kinetics
Automation ready—easy automation for high-throughput needs on the Kingfisher Purification System

Consistency and reproducibility  
The monolayer of streptavidin leaves most biotin-binding sites available for the binding of free biotin and biotinylated ligands. The specific and defined surface allows for efficient capture, separation, and downstream handling. In addition, the low size distribution (<5%) of the monosized beads and the streptavidin monolayer ensure negligible leakage, providing batch consistency and reproducibility of results.

High binding capacity 
The size of the ligand and the biotinylation procedure will affect the binding capacity. The capacity also depends on steric availability and charge interaction between bead and ligand and between ligands. With two or three biotin binding sites available for each streptavidin molecule on the surface of the bead after immobilization, one mg of Dynabeads M-280 Streptavidin typically binds: 
• 490-750 pmol free biotin
• 10 μg biotinylated IgG
• ∼200 pmol biotinylated peptides
• ∼10 μg biotinylated ds-DNA
• ∼200 pmol biotinylated ss-oligonucleotides

Applications
Thermo Fisher streptavidin-coupled Dynabeads magnetic beads have been used and cited for a wide variety of applications. Although the beads can be used to isolate any biotinylated targets, these hydrophobic beads are particularly well suited for immunoassays, protein purification, cell isolation, phage display, and biopanning.

Automation ready with the KingFisher Purification System
The 2.8-μm magnetic beads are ideal for high-throughput enrichment. This process can be automated using any of our KingFisher Sample Purification Systems or liquid handlers.

Commercial supply
Our manufacturing sites are ISO 13485-certified. Bead characteristics and manufacturing conditions help ensure consistency and batch reproducibility making them an ideal choice for commercial supply. If you are in the process of customizing Dynabeads M-280 Streptavidin beads in a commercial product or service or need a larger bulk volume of this product, please contact us at oemdynal@lifetech.com or see our Dynabeads OEM page.

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY
Specifications

Specifications

650-900 pmoles/mg beads
10 mg/mL
Brown
Magnetic Polystyrene
Beads in suspension
High-throughput Compatible
Streptavidin
Any Sample Type
Tosylactivated, Hydrophobic
2 mL
For Research Use Only
ISO9001 and ISO13485
Dynabeads M-280 Streptavidin is supplied in PBS, pH 7.4 with 0.02% NaN3. Store at 2°C to 8°C.

Capture of Biotinylated Targets
Streptavidin Magnetic Beads
KingFisher™ Sample Purification System, DynaMag™ magnets
Magnetic Beads
Dynabeads™
Ambient Temperature
Monosized 2.8 μm (CV <5%)
2.8 μm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.