missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Dynabeads™ MyOne™ Tosylactivated

When you wish to covalently couple antibodies, peptides, intact proteins, and functional enzymes to the surface of a magnetic bead choose Dynabeads Tosylactivated.

892.00 € - 3230.00 €

Specifications

Content And Storage Store at 2°C to 8°C.
Product Type Tosylactivated Bead
Elution Volume Min 5 μL
Format Beads in solution
High-throughput Compatibility High-throughput Compatible
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10466863
View Documents
Invitrogen™
65501
2 mL
892.00 €
Each
In Stock
to see stock.
Add to basket
 
10513375
View Documents
Invitrogen™
65502
10 mL
3230.00 €
Each
Estimated Shipment: 13-12-2024
to see stock.
Add to basket
 
Description

Description

When you wish to covalently couple antibodies, peptides, intact proteins, and functional enzymes to the surface of a magnetic bead choose Dynabeads Tosylactivated. Optimal antibody orientation makes these beads an excellent choice for the immunoprecipitation of proteins and protein complexes.

Ideal for Immunoprecipitation of protein complexes:
Low background and covalent linking of antibodies to the bead surface make Dynabeads Tosylactivated an excellent choice for Immunoprecipitation of proteins and protein complexes (Co-Immunoprecipitation, Co-IP). Gentle and rapid magnetic concentration of the beads and short incubation times (possible due to fast surface-based binding kinetics) make Dynabeads Tosylactivated an excellent choice for the immunoprecipitation of highly-labile or and/or transient (short-lived) protein complexes.

Bead Surface Characteristics for Dynabeads Tosylactivated:
• p-toluene-sulfonyl (Tosyl) groups
• Hydrophobic, pH Neutral
• Covalent binding by primary amine (NH2) or sulphydryl (SH) groups

Ultra rapid protocols:
• Identify members of protein complexes in minutes rather than hours
• Temporal resolution short enough to identify transient and labile complexes
• Identify binding partners that cannot be identified with longer protocols
• Rapid protocols further reduce the already ultra-low background binding
• Stronger signal to noise-ratios

Applications:
• IP of proteins and protein complexes
• Couple functional enzymes to bead surface for downstream assays
• Couple peptides to bead surface to identify binders

Purify peptides, proteins and enzymes that are:
• Transiently stable
• Structurally intact
• Temperature labile
• In their native conformation and functional

Coupling procedure outline:
Covalent coupling is performed overnight by incubating the desired ligand with the Dynabeads Tosylactivated. Ligands commonly coupled to Dynabeads Tosylactivated include peptides and proteins (e.g. antibodies for Immunoprecipitation or Co-Immunoprecipitation). Coupling occurs at neutral to high pH and at 37°C. We recommend coupling at pH 8.5-9.5, but for pH labile ligands, coupling can be performed in an alternative buffer at pH 7.4.

Upon completion of the ligand coupling step, the actual Dynabeads Tosylactivated surface coating will be rendered inert, resulting in low non-specific binding.

Binding capacity per milligram beads:
Varies depending on ligand, (e.g. 14-19 μg IgG)

About Dynabeads:

Dynabeads are non-porous monodisperse superparamagnetic beads. They are highly mobile in solution enabling ligands coupled to the beads to continuously interact with the entire sample volume. The superparamagnetic beads are pulled to the tube walls by transferring the tube to a rack containing a strong magnetic field. Strong magnetic fields quickly pull the beads to the tube wall, allowing for easy and complete removal of the supernatant by pipette. Washing steps are performed similarly.

Order Info

Shipping Condition: Room Temperature

Specifications

Specifications

Store at 2°C to 8°C.
5 μL
High-throughput Compatible
Research Grade
Room Temperature
Tosylactivated Bead
Beads in solution
DYNAL™, Dynabeads™, MyOne™
Any Sample Type
p-Toluene-Sulfonyl, Tosylactivated
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only. Not for use in diagnostic procedures.