missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SampleRack (for DynaMag™-2 magnet)

This removable plastic rack supplied with the DynaMag-2 magnet.

Brand:  Invitrogen™ 12322D

120.65 EUR valid until 2025-06-30
Use promo code "24074" to get your promotional price.


Product Code. 11350972

  • 160.00 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket

Includes:

One acrylic copolymer SampleRack. Holds 16 microcentrifuge tubes.

This item is not returnable. View return policy

Description

Description

This removable plastic rack supplied with the DynaMag-2 magnet

  • Holds 16 x 1.5 mL micro-centrifuge tubes
  • Tubes are kept in SampleRack, before, during and after isolation
  • When handling multiple samples in parallel, use additional SampleRacks to increase capacity
  • With two racks, keep up to 16 tubes in same SampleRack throughout Dynabeads isolation procedure, moving rack on and off the magnet as needed
  • SampleRack can be placed on bench, in refrigerator or incubator

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Sample Rack
DynaMag™-2
Each
Contains one acrylic copolymer SampleRack. Holds 16 microcentrifuge tubes.

Store at 4-30°C.
DynaMag™
Room Temperature
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Manuals

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ SampleRack (for DynaMag™-2 magnet) > 1 Ea.

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.