missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ E-Gel™ Sample Loading Buffer, 1X
GREENER_CHOICE

Supplied as a ready-to-use 1X solution

Brand:  Invitrogen™ 10482055

54.63 EUR valid until 2025-02-28
Use promo code "24154" to get your promotional price.


Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Product Code. 10112098

  • 58.75 € / Pack of 4
In Stock
to see stock.
Edge
Explore more special offers
Add to basket

Includes: E-Gel™ Sample Loading Buffer is supplied as a 1X solution in buffer containing 0.0094% XCFF and 0.06% tartrazine
This item is not returnable. View return policy

Description

Description

E-Gel™ Sample Buffer is formulated specifically for maximum performance on E-Gel EX gels as well as other E-Gel precast gels.

Agarose Gel Electrophoresis, DNA and RNA Purification and Analysis, Nucleic Acid Gel Electrophoresis and Blotting

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY
Specifications

Specifications

• 4 x 1.25 mL 1X E-Gel Sample Loading buffer

Store at room temperature or at 4°C for up to 6 months. For longer periods, store at -20°C.

1X
Tartrazine, Xylene Cyanol, Xylene Cyanol FF (XCFF)
1.25 mL
E-Gel™ agarose gels
E-Gel™
Approved for shipment at Room Temperature or on Dry Ice
Sample Loading Buffers
Agarose
Sample Loading Buffer
Liquid
Sustainable packaging
4 x 1.25 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.