missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ One Shot™ TOP10 Electrocomp™ E. coli

Product Code. 10423792
Click to view available options
Quantity:
11 x 50 μL, 10 Reactions
21 x 50 μL, 20 Reactions
Unit Size:
20 reactions
Each
This item is not returnable. View return policy

Product Code. 10423792

Brand: Invitrogen™ C404050

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

One Shot TOP10 Electrocomp E. coli cells are ideal for high-efficiency cloning and plasmid propagation. Provided at a transformation efficiency of >1 x 1010 cfu/ μg supercoiled DNA, these cells allow for stable replication of high copy number plasmids.

One Shot TOP10 Electrocomp E. coli cells are ideal for high-efficiency cloning and plasmid propagation. Provided at a transformation efficiency of >1 x 1010 cfu/ μg supercoiled DNA, these cells allow for stable replication of high copy number plasmids. TOP10 Electrocomp E. coli cells are transformed via electroporation and achieve higher transformation efficiencies than chemically competent cells. They can therefore be used in more challenging applications, including cloning low amounts of DNA or cDNA and gDNA library construction.

TOP10 E. coli cells are genetically similar to the DH10B strain but have been reported to be more resilient to stress conditions like osmotic shock and acidic pH stress. These cells carry mutations in the methylation-dependent restriction system (mcrA, mcrBC, and mrr) allowing them to clone both prokaryotic and eukaryotic genomic DNA, as well as efficient plasmid rescue from eukaryotic genomes. Similar to other DH strains, TOP10 has the lacZΔM15 genotype, providing for the option of blue-white screening on plates containing either X-Gal or Bluo-Gal.

One Shot TOP10 Electrocomp E. coli cells offer:

  • Transformation efficiencies of >1 x 1010 cfu/μg
  • hsdR for efficient transformation of unmethylated DNA from PCR amplifications
  • mcrA for efficient transformation of methylated DNA from genomic preparations
  • lacZΔM15 for blue/white color screening of recombinant clones
  • endA1 for cleaner DNA preparations and better results in downstream applications due to elimination of nonspecific digestion by Endonuclease I
  • recA1 for reduced occurrence of nonspecific recombination in cloned DNA

Easy-to-use One Shot format

TOP10 Chemically Competent E. coli cells are supplied in the convenient, single-reaction One Shot format. The single-tube, single-use format allows all steps of the transformation protocol, up to plating, to take place in the same tube, thereby helping save time and prevent contamination.

Genotype

FmcrA Δ(mrr-hsdRMS-mcrBC) φ80lacZΔM15 ΔlacX74 recA1 araD139 Δ(ara-leu)7697 galU galK λ–rpsL(StrR) endA1 nupG

Find the strain and format that fit your needs

We offer other DH strains in chemically competent and electrocompetent cell formats.

Electroporation cuvettes with 0.1 cm (Cat. No. P41050), 0.2 cm (Cat. No. P45050), and 0.4 cm (Cat. No. P46050) gaps are available.

The TOP10 strain is available in several MultiShot formats for high throughput applications.

Explore bacterial growth media formats.

Note: TOP10 cells do not require IPTG to induce expression from the lac promoter. A high-voltage electroporation apparatus is required.

Order Info

Shipping Condition: Dry Ice

TRUSTED_SUSTAINABILITY

Specifications

Product Type Electrocompetent Cells
Contains F' Episome No
Improves Plasmid Quality Yes (endA1)
Cloning Methylated DNA Yes (mcrA)
Transformation Efficiency Level High Efficiency (>1 x 1010 cfu/μg)
Content And Storage • One Shot TOP10 Electrocomp E. coli (11 x 50 μL)
Store at –80°C.

• pUC19 DNA (50 μL)
Store at –20°C.

• S.O.C. Medium (6 mL)
Store at room temperature.

Antibiotic Resistance Bacterial Yes (Streptomycin)
Cloning Unstable DNA No
Blue/White Screening Yes (lacZΔM15)
High-throughput Compatibility Low
Plasmid May be used for plasmids >20 kb
Preparing Unmethylated DNA No
Reduces Recombination Yes (recA13)
Shipping Condition Dry Ice
T1 Phage - Resistant (tonA) No
Species E. coli (K12)
Format Tube
Product Line One Shot
Quantity 11 x 50 μL, 10 Reactions
Mehr anzeigen Weniger anzeigen

For Research Use Only. Not for use in diagnostic procedures.

Berichtigung von Produktinhalten

Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.

Name des Produkts

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.

Vielen Dank, dass Sie uns helfen, unsere Website zu verbessern. Ihr Feedback wurde übermittelt