Learn More
Invitrogen™ FreeStyle™ CHO-S Cells
Rapid and high-yield mammalian protein production
Brand: Invitrogen™ R80007
837.46 EUR valid until 2025-02-21
Use promo code "24115" to get your promotional price.
Description
Animal origin-free, chemically defined, low-protein, reduced growth factors, reduced serum, xeno-free. The FreeStyle MAX System is a breakthrough technology for rapid and high-yield mammalian protein production. This new mammalian expression technology is available in two convenient formats. FreeStyle MAX Reagent is a novel cationic lipid-based reagent designed to transfect FreeStyle cells with high efficiency. The FreeStyle MAX Expression System combines GIBCO™ FreeStyle Media, FreeStyle MAX Reagent, and either FreeStyle CHO-S cells or FreeStyle 293 cells. The only equipment required to use the FreeStyle MAX System is a CO2 incubator and a platform shaker with shake flasks or a spinner culture.
- FreeStyle MAX System provides:An easy, rapid protocol for transient transfection of CHO and 293 cells in suspension cultures
- Functional proteins (up to milligram quantities) with all post-translational modifications produced in one week
- Simplified downstream purification of secreted proteins with FreeStyle serum-free medium
- Straightforward protocol for production scale-up
- Replace time-consuming stable cell line generation with large-scale functional protein production in one week
Antibody and Cell Culture Production, Bioproduction, Cell Culture, Mammalian Cell Culture, Mammalian Expression, Mammalian Expression Cell Lines, Mammalian Protein Production, Plasmid Transfection, Protein Expression, Proteins, Expression, Isolation and Analysis, Transfection, Viral Vaccine Production
Specifications
FreeStyle™ CHO-S Cells | |
CHO-S | |
Hamster | |
Freestyle™ | |
1 mL | |
FreeStyle™ |
1 vial of 1 x 107 cells. Store in liquid nitrogen. | |
Lipid-based Transfection | |
cand Transfection | |
CHO-S Cell | |
1 x 107 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.