Learn More
Invitrogen™ FreeStyle™ MAX CHO Expression System
Rapid and high-yield mammalian protein production
Brand: Invitrogen™ K900020
1495.78 EUR valid until 2025-02-21
Use promo code "24115" to get your promotional price.
Description
FreeStyle MAX Reagent is a novel cationic lipid-based reagent designed to transfect FreeStyle cells with high efficiency. The FreeStyle MAX Expression System combines GIBCO™ FreeStyle Media, FreeStyle MAX Reagent, and either FreeStyle CHO-S cells or FreeStyle 293 cells. The only equipment required to use the FreeStyle MAX System is a CO2 incubator and a platform shaker with shake flasks or a spinner culture.
- Easy, rapid protocol for transient transfection of CHO and 293 cells in suspension cultures
- Functional proteins (up to milligram quantities) with all post-translational modifications produced in one week
- Simplified downstream purification of secreted proteins with FreeStyle serum-free medium
- Straightforward protocol for production scale-up
- Replace time-consuming stable cell line generation with large-scale functional protein production in one week
Antibody and Cell Culture Production, Bioproduction, Cell Culture, Mammalian Expression, Mammalian Protein Production, Plasmid Transfection, Protein Expression, Proteins, Expression, Isolation and Analysis, Transfection, Transient Protein Production and High-Throughput Screening
Specifications
FreeStyle™ CHO-S Cells | |
Store FreeStyle cells in liquid nitrogen. Store FreeStyle MAX Reagent at 2–8°C. Store FreeStyle CHO Expression Medium, FreeStyle 293 Expression Medium, and OptiPRO SFM at 2–8°C in the dark. Store the pCMV SPORT at -20°C. Stability is guaranteed for 6 months under the recommended storage conditions. Please refer to the manual for additional storage information. | |
Suspension Cell Culture | |
Lipid-based Transfection | |
Mammalian | |
Protein Production | |
Expression System |
Animal Origin-Free, Serum-Free | |
Transfection | |
CHO | |
Cell-Based Expression | |
Protein Expression | |
Freestyle™ | |
1 kit |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.