Learn More
Invitrogen™ pcDNA™6.2/V5-PL-DEST Mammalian Expression Vector
Designed for high-level, constitutive expression in variety of mammalian cell lines
Brand:  Invitrogen™ 12537162
370.68 EUR valid until 2025-03-21
Use promo code "24111" to get your promotional price.
Product Code. 11578706
  • 556.00 € / 6µg
Explore more special offers
Alert:
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24111” to get your promotional price
Includes: All destination vectors are provided lyophilized and supercoiled.
Requires : Gateway™ entry clone, appropriate Gateway™ LR Clonase enzyme mix, and reaction buffer.
Description
Description
  • Promoterless version of pcDNA™6.2/V5-DEST vector
  • attR sites for Gateway cloning
  • Compatible with MultiSite Gateway Pro kits
  • C-terminal V5 tag for easy detection
  • Blasticidin resistance gene for efficient stable selection
  • Ampicillin resistance gene and pUC origin for selection and maintenance in E. coli
  • Gateway Cloning Destination vectors have attR sites for recombination with any attL-flanked fragment, regardless of whether it is an entry clone or an UltimateORF Clone
Cloning,Constitutive Expression,Gateway Cloning,Mammalian Expression,Multi-Gene Expression,Multisite Gateway,Protein Expression,Proteins, Expression, Isolation and Analysis
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Transfection
None (Promoterless)
All destination vectors are provided lyophilized and supercoiled.
C-terminal
Gateway
pcDNA, pDEST, MultiSite Gateway Vectors
Clone your own Promoter, Multi-Gene Expression
MultiSite Gateway™ Pro kits (e.g. cat. no. 12537-100)
Mammalian Expression Vector
Mammalian
V5 Epitope Tag
6 μg
Gateway™, pcDNA™