missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ pENTR™/D-TOPO™ Cloning Kit, with One Shot™ Mach1™-T1R Chemically Competent E. coli
Click to view available options
Quantity:
20 Reactions
Unit Size:
Each
Description
Includes
Ready-to-use TOPO vector, dNTPs, salt solution, sterile water, universal M13 sequencing primers, TOP10 or Mach1-T1R Chemically E. coli, S.O.C. Medium, and a pUC19 control plasmid.
- Blunt-end PCR products clone directionally at greater than 90% efficiency, with no ligase, post-PCR procedures, or restriction enzymes required
- Rapidly shuttle cloned genes between multiple vector systems
- Reliable performance
- For access to Gateway System, PCR amplify gene of interest and add product straight to provided topoisomerase charged pENTR/D-TOPO vector, incubate 5 minutes, and transform provided competent E. coli cells
- Resulting attL containing Gateway Entry clones are ready for efficient recombination with choice of Gateway Destination vectors
- pENTR/D-TOPO vector includes M13 and T7 primer sequencing sites and attL recombination sites flanking PCR product insertion site., allowing clones to be easily sequence-verified and recombined into choice of attR containing Gateway destination vectors
- Kanamycin resistance gene and pUC origin are used for selection and high-copy propagation in E. coli
- Four base overhang on vector pairs with four base sequence designed into forward primer used in PCR reaction to provide directionality to the topoisomerase ligation reaction
- Move same sequence of DNA between different vectors without using restriction enzymes, ligase, subcloning steps, screening of countless colonies, or resequencing
Cloning/Gateway Cloning
Specifications
Specifications
| Cloning Method | Directional TOPO™ |
| Content And Storage | Each Directional TOPO™ pENTR Cloning Kit contains the appropriate, ready-to-use TOPO™ vector, dNTPs, salt solution, sterile water, universal M13 sequencing primers, TOP10 or Mach1-T1R Chemically E. coli, S.O.C. Medium, and a pUC19 control plasmid. Store Competent E. coli at -80°C. Store all other components at -20°C. |
| No. of Reactions | 20 reactions |
| Product Line | One Shot |
| Product Type | TOPO Cloning Kit |
| Quantity | 20 Reactions |
| Vector | pENTR |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction