missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ pENTR™/D-TOPO™ Cloning Kit, with One Shot™ TOP10 Chemically Competent E. coli
Efficient, 5-minute cloning strategy to directionally clone blunt-end PCR product into vector for entry into Gateway System or MultiSite Gateway System
Brand: Invitrogen™ K240020
664.64 EUR valid until 2025-04-18
Use promo code "24107" to get your promotional price.
Includes: pENTR/D-TOPO vector, dNTPs, salt solution, sterile water, universal M13 sequencing primers, OneShot TOP10 Chemically E. coli, S.O.C. Medium, and a pUC19 control plasmid.
Description
- Blunt-end PCR products clone directionally at greater than 90% efficiency, with no ligase, post-PCR procedures, or restriction enzymes required
- Rapidly shuttle cloned genes between multiple vector systems
- Reliable performance
- Directional cloning vector for entry to Gateway System
- For access to Gateway System, PCR amplify gene of interest and add product straight to provided topoisomerase charged pENTR/D-TOPO vector, incubate 5 minutes, and transform provided competent E. coli cells
- Resulting attL containing Gateway Entry clones are ready for efficient recombination with the choice of Gateway Destination vectors
- The pENTR/D-TOPO vector includes M13 and T7 primer sequencing sites and attL recombination sites flanking the PCR product insertion site, allowing clones to be easily sequence-verified and recombined into the choice of attR containing Gateway destination vectors
- Kanamycin resistance gene and a pUC origin are used for selection and high-copy propagation in E. coli
- Four base overhang on vector pairs with four base sequence designed into forward primer used in PCR reaction to provide directionality to the topoisomerase ligation reaction
- Circumvents the limitations of restriction mediated cloning, enabling to access virtually any expression system in a simple one hour, 99%-efficient and reversible, Gateway recombination reaction
- Move same sequence of DNA between different vectors without using restriction enzymes, ligase, subcloning steps, screening of countless colonies, or resequencing to save time, money, and effort
Cloning, Gateway Cloning

Specifications
Directional TOPO™ | |
20 reactions | |
TOPO Cloning Kit | |
pENTR |
pENTR™⁄D-TOPO™ Cloning Kit contains pENTR⁄D-TOPO™ vector, dNTPs, salt solution, sterile water, universal M13 sequencing primers, OneShot™ TOP10 Chemically E. coli, S.O.C. Medium, and a pUC19 control plasmid. Store Competent E. coli at -80°C. Store all other components at -20°C. | |
One Shot™ | |
20 Reactions |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Invitrogen™ pENTR™/D-TOPO™ Cloning Kit, with One Shot™ TOP10 Chemically Competent E. coli > 20 reactions
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?Share a Content Correction