missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Gateway™ Vector Conversion System with One Shot™ ccdB Survival Cells
Click to view available options
Quantity:
1 kit
Unit Size:
1 set
Description
Includes
Gateway Reading Frame Cassettes A, B, and C.1, pENTR™-gus positive control, and ccdB Survival Cells.
- Gateway cassette containing attR recombination sites flanking ccdB gene (1) and chloramphenicol-resistance gene are blunt-end cloned into multiple cloning site of any vector ccdB
- Survival 2 competent cells allow propagation of Gateway vectors containing ccdB gene
- Three cassettes to construct a destination vector in correct reading frame; Each cassette has unique restriction site to easily distinguish cassettes and screen for correct orientation
Cloning,Gateway Cloning,Gateway Vector Conversion Systems
Specifications
Specifications
| Product Type | Vector Conversion System |
| Content And Storage | The Gateway™ Vector Conversion System contains Gateway™ Reading Frame Cassettes A, B, and C.1, pENTR™-gus positive control, and One Shot™ ccdB Survival Cells. Store DNA at -20°C and competent cells at -80°C. |
| Antibiotic Resistance Bacterial | Chloramphenicol (CmR) |
| Bacterial or Yeast Strain | ccdB Survival™ 2 |
| Cloning Method | Gateway |
| Quantity | 1 kit |
| Product Line | One Shot |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction