missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Taq DNA Polymerase, recombinant

Taq DNA Polymerase is a thermostable enzyme that synthesizes DNA from single-stranded templates in the presence of dNTPs and a primer.

Brand:  Invitrogen™ 10342020

 View more versions of this product

Product Code. 10033382

  • 267.00 € / 500 units

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

Includes: Taq DNA Polymerase (5U/μL); 10X PCR buffer (200mM Tris-HCl pH 8.4, 500mM KCl); Magnesium Chloride (50mM)
This item is not returnable. View return policy

Description

Description

Taq DNA Polymerase, recombinant, is a thermostable enzyme that synthesizes DNA from single-stranded templates in the presence of dNTPs and a primer. This recombinant enzyme is purified from the cloned Thermus aquaticus DNA polymerase gene expressed in E. coli and consists of a single polypeptide with a molecular weight of 94 kDa. It has a 5' to 3' DNA polymerase activity and a 5' to 3' exonuclease activity.

Features

  • A choice of recombinant or native enzyme
  • Amplification of PCR products up to 5 kb in size
  • Enzyme is licensed and qualified for PCR

Applications

  • Amplification of DNA from complex genomic, viral, and plasmid templates
  • RT-PCR
  • Sequencing ssDNA
  • Cycle sequencing

Notes

  • One unit of Taq DNA Polymerase is the amount of enzyme required to incorporate 10 nmoles of deoxyribonucleotide into DNA in 30 minutes at 74°C.
  • For superior PCR performance, DreamTaq DNA Polymerase is recommended.

Order Info

Shipping Condition: Approved for shipment on wet or dry ice

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Taq DNA Polymerase (5 U/μL), 100 μL
• 10X PCR buffer, 2.5 mL
• 50 mM MgCl2, 1 mL

Store at -10°C to -30°C in a non-frost-free freezer.
Guaranteed stable for 6 months when stored properly.
Low
Taq DNA Polymerase
5' - 3'
500 Units
Standard PCR
1X
500 Reactions
Separate Components
DNA
Stand-alone enzyme
qPCR, Standard PCR
Standard
DNA Polymerase
Dry Ice
5 U/μL
No
3'-A
5 kb or less
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.