missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ imMedia™ Growth Medium, agar, ampicillin, X-gal/IPTG

Microwaveable, premixed, presterilized E. coli growth medium that can be prepared in just five minutes without autoclaving

Brand:  Invitrogen™ Q60220

Product Code. 10308022

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers

Includes: imMedia growth medium is provided in a 20-pouch box. Each pouch contains sufficient reagents to prepare 200mL of liquid medium or 8-10 agar plates.
This item is not returnable. View return policy

Description

Description

  • Contains everything needed to prepare low-salt LB medium or agar plates, saving hours of medium preparation time
  • No weighing or mixing of media components
  • No autoclaving
  • No waiting for media to cool before adding antibiotics

Bioproduction, Cloning, Transformation

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Ampicillin
Powder
Microwave
E. coli
Ampicillin
Ready-to-Use Individual Pouches
Each type of imMedia™ growth medium is provided in a 20-pouch box. Each pouch contains sufficient reagents to prepare 8-10 standard 100 mm2 agar plates.

Store at room temperature.

All reagents are guaranteed stable for 12 months when properly stored.

LB, Quick-Preparation Media (e.g. Immedia™)
Bacterial Growth Medium
Agar Plates
Powder Reagents
imMedia™
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.