missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ LB Broth Base (Lennox)

LB Broth Base (Lennox formulation) is a popular, rich medium used to cultivate bacteria and perform coliphage plaque assays.

Brand:  Invitrogen™ 12780029

 View more versions of this product

Product Code. 11528866

  • 322.00 € / Each
Estimated Shipment: 29-10-2024
to see stock.
Edge
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

LB Broth Base (Lennox formulation) is a popular, rich medium used to cultivate bacteria and perform coliphage plaque assays. The original formulation was published in 1951 by Guiseppe Bertani in his study of phage plaque formation and named Lysogeny broth. It is also known as Lennox broth and Luria-Bertani broth.

Widely used LB medium formulations differ in salt concentration:
• Miller: 10 g/L NaCl
• Lennox: 5 g/L NaCl
• Luria: 0.5 g/L NaCl

LB Broth Base is recommended for the general maintenance and growth of E. coli as it allows for fast cell expansion and reach high cell density. Its Lennox formulation has lower levels of salts than Miller and is recommended to use with salt-sensitive antibiotics such as Zeocin, tetracycline, or chloramphenicol.

LB Broth Base formulation and preparation
To prepare 1 L of Lennox-LB media, weigh 20 g of powder, and add distilled water to 1 L. Adjust the pH value to 7.0 with 5 N NaOH and sterilize by autoclaving for 25 minutes at 121°C. Formulation per liter: 10 g SELECT Peptone 140, 5 g SELECT Yeast Extract, and 5 g NaCl. 

Primary applications
• Growth and maintenance of E. coli strains for plasmid purification and/or protein expression
• Resuspension of T. pyriformis cell fractions

Find the media format that fits your needs
We offer other ready-to-use media formulations, such as LB (Lennox) (Cat. No. 10855001), Terrific Broth (Cat. No. A1374301) or S.O.C recovery medium (Cat. No. 15544034).

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Escherichia coli
Bottle
Autoclave
Coliforms
Room Temperature
• 2.5 kg Dehydrated Media Powder

Store at room temperature.

LB Broth Base (Lennox L Broth Base)
Dehydrated Culture Media
2.5 kg
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.