Learn More
Invitrogen™ Lipofectamine™ 2000 Transfection Reagent
GREENER_CHOICE
Versatile transfection reagent that has been shown to effectively transfect the widest variety of adherent and suspension cell lines
Brand:  Invitrogen™ 11668030
245.43 EUR valid until 2024-12-31
Use promo code "21651" to get your promotional price.
 View more versions of this product
Product Code. 12313563
  • 303.00 € / Each
Edge
Explore more special offers
Description
Description
Lipofectamine™ 2000 Transfection Reagent is a versatile transfection reagent that has been shown to effectively transfect the widest variety of adherent and suspension cell lines. Researchers use Lipofectamine™ 2000 Reagent for siRNA- and shRNA-based gene knockdown experiments, as well as for gene expression studies. Lipofectamine™ 2000 Transfection Reagent is our best transfection reagent available for the co-transfection of siRNA and plasmid DNA.
With Lipofectamine™ 2000 Transfection Reagent, you'll get:
  • Exceptional transfection efficiency in the broadest range of cell lines and the highest levels of recombinant protein expression (see table)
  • Superior performance for co-transfection of siRNA and plasmid DNA
  • Proven efficacy in the presence of serum—eliminates the need to change media following transfection
  • Reliable performance for high-throughput applications
  • The best choice for establishing stable cell lines
    A high-performance transfection reagent for gene expression and gene silencing
    Lipofectamine™ 2000 Transfection Reagent works effectively with all common cell lines as well as with many challenging ones, and can be used in media with or without serum. For gene silencing, Lipofectamine™ 2000 Transfection Reagent's high-efficiency transfections provide the high levels of gene knockdown needed to achieve convincing results. Lipofectamine™ 2000 Transfection Reagent is the number one choice for co-transfection, given its effectiveness for transfecting both siRNA and plasmid DNA. Lipofectamine™ 2000 Transfection Reagent is easy to use—simply mix with nucleic acid and add to cell culture.
    Ideal for high-throughput work
    Simplicity and speed combined with high transfection efficiency make Lipofectamine™ 2000 Transfection Reagent ideal for transient protein expression or high-throughput RNAi transfections. Transfection conditions can be easily established for automated or robotic systems used in such applications.
    Order Info
    Sustainable packaging. Approved for shipment at Room Temperature or on Wet Ice.
    TRUSTED_SUSTAINABILITY
  • Specifications
    Specifications
    Each unit contains one vial of Lipofectamine™ 2000 Reagent (0.3 ml). Store at 2–8°C.
    12-well Plate
    Transfection
    High-throughput Compatible
    Transfection Reagent
    Yes
    Lipid-Based Transfection
    Established Cell Lines, Stem Cells, Primary Cells, Hard-to-Transfect Cells
    Plasmid DNA, Synthetic siRNA, RNAi Plasmids (shRNA, miR)
    Sustainable packaging
    Lipofectamine™
    0.3 mL
    Approved for shipment at Room Temperature or on Wet Ice