missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Lipofectamine™ RNAiMAX Transfection Reagent
![GREENER_CHOICE](/content/dam/fishersci/glyphs/green_glyph.png)
Provides highest transfection efficiencies on widest variety of cell types for siRNA-mediated gene knockdown experiments
Brand: Invitrogen™ 13778150
1170.40 EUR valid until 2025-03-28
Use promo code "24107" to get your promotional price.
This item is not returnable.
View return policy
Description
Lipofectamine™ RNAiMAX Transfection Reagent provides the highest transfection efficiencies on the widest variety of cell types for siRNA-mediated gene knockdown experiments. Lipofectamine™ RNAiMAX is a proprietary RNAi-specific cationic lipid formulation designed specifically for the delivery of siRNA and miRNA into all cell types.With Lipofectamine™ RNAiMAX Transfection Reagent you will get:
High knockdown in a wide range of cells
Lipofectamine™ RNAiMAX Transfection Reagent transfects a wide range of cell types (see figure). For gene silencing, Lipofectamine™ RNAiMAX Transfection Reagent's high-efficiency transfections lead to the high levels of gene knockdown needed to achieve convincing results.
Simple, high-throughput–ready transfections
Simply mix Lipofectamine™ RNAiMAX Transfection Reagent with siRNA, add to your cells, incubate, and measure gene knockdown. The simplicity and speed combined with high transfection efficiency make Lipofectamine™ RNAiMAX Transfection Reagent ideal for high-throughput siRNA transfections. Transfection conditions can be easily established for automated or robotic systems used in such applications.
Find an optimized Lipofectamine™ RNAiMAX transfection protocol for your cell line.
Order Info
Shipping Condition: Qualified for Ambient or Wet Ice
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Contains one vial (1.5 ml) Lipofectamine™ RNAiMAX Transfection Reagent. Store at 2–8°C. Do not freeze. | |
6-well Plate, 12-well Plate, 24-well Plate, 48-well Plate, 96-well Plate, Flasks | |
Lipid-based Transfection | |
Sustainable packaging | |
Lipofectamine™ | |
1.5 mL | |
Approved for shipment at Room Temperature or on Wet Ice |
Established Cell Lines, Stem Cells, Primary Cells, Hard-to-Transfect Cells | |
siRNA, Synthetic siRNA, RNAi Plasmids (shRNA, miR), RNAi, miRNA | |
Transfection | |
High-throughput Compatible | |
Transfection Reagent | |
Yes |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?Share a Content Correction