missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ M-MLV Reverse Transcriptase (200 U/μL)

M-MLV Reverse Transcriptase is a recombinant DNA polymerase that synthesizes a complementary DNA strand from single-stranded RNA, DNA, or an RNA:DNA hybrid.

Brand:  Invitrogen™ 28025021

 View more versions of this product

Product Code. 10247352

  • 1810.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

M-MLV Reverse Transcriptase is a recombinant DNA polymerase that synthesizes a complementary DNA strand from single-stranded RNA, DNA, or an RNA:DNA hybrid. Compared to AMV RT, Moloney Murine Leukemia Virus Reverse Transcriptase (M-MLV RT) lacks DNA endonuclease activity and has a lower RNase H activity.

One unit of M-MLV RT is the amount of enzyme required to incorporate 1 nmole of deoxyribonucleotide into acid-precipitable material in 10 min at 37°C using poly(A) oligo(dT)25 as template-primer.

Features of M-MLV Reverse Transcriptase include:
• Excellent thermostability with optimal activity at 37°C
• Can be used to synthesize first-strand cDNA up to 7 kb
• Multiple applications including synthesis of first-strand cDNA, primer extension, sequencing dsDNA, cDNA libraries, and RT-PCR

Source
Purified from E. coli expressing the pol gene of M-MLV on a plasmid

Performance and quality testing
SDS-PAGE purity; endodeoxyribonuclease, exodeoxyribonuclease, and ribonuclease assays; and yield and length of cDNA product

Unit reaction conditions
50 mM Tris-HCl (pH 8.3), 40 mM KCl, 6 mM MgCl2, 1 mM DTT, 0.5 mM [3H]dTTP, 0.1 mM poly(A), 0.1 mM oligo(dT)25, 0.1 mg/mL BSA, and enzyme in 50 μL for 10 min at 37°C

Order Info

Shipping Condition: Wet ice

TRUSTED_SUSTAINABILITY
Specifications

Specifications

• M-mLV RT, 1 mL (200 U/μL)
• First Strand Buffer, 4 x 1 mL
• DTT, 4 x 500 μL (100 mM)

Store at –20°C.

50 min.
37°C
M-MLV
Wet Ice
First-Strand cDNA
Separate components
Up to 7 kb
Stand-alone Enzyme
Reverse Transcription
200,000 units
Yes
200 U/μL
1,000 Reactions
Reverse Transcription
RNA
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.