missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ M-MLV Reverse Transcriptase Buffer

M-MLV Reverse Transcriptase Buffer is for use with Moloney Murine Leukemia Virus Reverse Transcriptase (M-MLV RT).

Brand:  Invitrogen™ 18057018

47.00 EUR valid until 2024-12-31
Use promo code "21651" to get your promotional price.


Product Code. 10512703

  • 58.75 € / 1mL

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers

Includes: 1mL M-MLV Reverse Transcriptase Buffer and 500μL DTT (0.1M)
This item is not returnable. View return policy

Description

Description

M-MLV Reverse Transcriptase Buffer is for use with Moloney Murine Leukemia Virus Reverse Transcriptase (M-MLV RT).

5X buffer composition
• 250 mM Tris-HCl (pH 8.3)
• 375 mM KCl
• 15 mM MgCl2

Included with
• M-MLV Reverse Transcriptase
• SuperScript Reverse Transcriptase
• SuperScript II Reverse Transcriptase
• SuperScript III Reverse Transcriptase

M-MLV Reverse Transcriptase Buffer does NOT contain actinomycin D, nucleotides, or dithiothreitol (DTT).

Order Info

Approved for shipment on wet or dry ice

TRUSTED_SUSTAINABILITY
Specifications

Specifications

• M-MLV Reverse Transcriptase Buffer (1 mL)
• 0.1 M DTT (500 μL)

Store at –20°C.

M-MLV Reverse Transcriptase
Wet or Dry Ice
Separate components
RNA
Reverse Transcription
1 mL
5X
Reverse Transcription
1 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.