missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ pcDNA™3.1/Zeo (+) Mammalian Expression Vector
Designed for high-level, constitutive expression in a variety of mammalian cell lines
Brand: Invitrogen™ V86020
712.80 EUR valid until 2024-09-27
Use promo code "21650" to get your promotional price.
Includes: 20μg of this pcDNA3.1/Zeo(+) vector, as well as an expression control, are supplied supercoiled and lyophilized.
Description
- Contains Zeocin selectable marker and forward-orientation multiple cloning site
- Cytomegalovirus (CMV) enhancer-promoter for high-level expression
- Large multiple cloning site in either forward (+) or reverse (-) orientations
- Bovine Growth Hormone (BGH) polyadenylation signal and transcription termination sequence for enhanced mRNA stability
- SV40 origin for episomal replication and simple vector rescue in cell lines expressing the large T antigen (i.e., COS-1 and COS-7)
- Ampicillin resistance gene and pUC origin for selection and maintenance in E. coli
Constitutive Expression, Mammalian Expression, Protein Expression, Proteins, Expression, Isolation and Analysis
Specifications
Constitutive | |
CMV | |
Zeocin™ | |
Untagged | |
20 μg | |
pcDNA™ |
Transfection | |
Mammalian Expression Vector | |
20 μg of this pcDNA™3.1/Zeo(+) vector, as well as an expression control, are supplied supercoiled and lyophilized. Store at -20°C. Vectors are guaranteed stable for 6 months when properly stored. | |
Restriction Enzyme/MCS | |
pcDNA | |
Constitutive Expression |
For Research Use Only. Not for use in diagnostic procedures.