Learn More
Invitrogen™ pGAPZ A, B, & C Pichia pastoris Expression Vectors
pGAPZ A, B, & C and pGAPZ A, B, & C are expression vectors designed for high-level, constitutive expression in Pichia pastoris.
Brand:  Invitrogen™ V20020
Product Code. 17144629
  • 1060.00 € / 20µg
Explore more special offers
Includes: All vectors are supercoiled and are provided lyophilized. A lacZ positive expression control strain is provided with the pGAPZ vector.
Description
Description
pGAPZ A, B, & C and pGAPZ A, B, & C are expression vectors designed for high-level, constitutive expression in Pichia pastoris. pGAPZ and pGAPZ were created by replacing the methanol-regulated AOX1 promoter with the constitutive, glyceraldehyde-3-phosphate dehydrogenase (GAP) promoter in the backbone of the pPICZ vectors. Although the yield of any protein constitutively expressed in the Pichia system is dependent on the toxicity of the protein to yeast, constitutive expression under the control of pGAPZ or pGAPZ vectors can sometimes produce greater yields than inducible expression (1).
TRUSTED_SUSTAINABILITY
Specifications
Specifications
GAP
All vectors are supercoiled and are provided lyophilized (store at -20°C). A lacZ positive expression control strain is provided with the pGAPZ vector.
Store strains at 4°C.
All reagents are guaranteed stable for 6 months when properly stored.
His Tag (6x), c-Myc Epitope Tag
20 μg
Pichia Expression Vector
Zeocin™ (ZeoR)
Restriction Enzyme/MCS
pGAP