Learn More
Invitrogen™ Platinum™ Taq DNA Polymerase
Invitrogen Platinum Taq DNA Polymerase is a convenient and reliable 'hot start' thermostable DNA polymerase for PCR that provides enhanced specificity over that of Taq DNA Polymerase.
Brand: Invitrogen™ 10966034
680.17 EUR valid until 2025-12-31
Use promo code "24357" to get your promotional price.
Includes: Each box contains: Platinum Taq DNA Polymerase; 10X PCR Buffer (no magnesium chloride); 50mM magnesium chloride; KB Extender
Description
Invitrogen Platinum Taq DNA Polymerase is a convenient and reliable hot-start thermostable DNA polymerase for PCR that provides enhanced specificity over that of Taq DNA Polymerase. The hot start property of the enzyme is conferred by thermolabile monoclonal antibodies that render Taq DNA polymerase inactive until the initial PCR denaturation step, thus preventing the extention of nonspecifically annealed primers and improving product yield.
The hot-start property allows for convenient reaction assembly at room temperature. Just as with Taq DNA Polymerase, Platinum Taq DNA Polymerase has a non-template dependent terminal transferase activity that adds a 3' deoxyadenosine to product ends and has a 5' to 3' exonuclease activity. PCR products generated with Platinum Taq DNA Polymerase may be used in the same downstream applications without protocol modifications.
Features
- Convenient reaction assembly at room temperature
- 5' to 3' exonuclease activity
- Lacks 3' to 5' exonuclease activity
Applications
- Amplification of DNA from complex genomic, viral, and plasmid templates
- Hot-start PCR
- RT-PCR
Notes
- For superior PCR performance, the next-generation enzyme Platinum II Taq Hot-start DNA Polymerase is recommended.
- Platinum II Taq Hot-Start DNA Polymerase is designed for universal primer annealing and fast, easy PCR with its unique combination of innovative buffer, high-performance engineered Taq DNA polymerase, and superior hot-start technology.
Order Info
Shipping Condition: Approved for shipment on wet or dry ice
Specifications
• Platinum Taq DNA Polymerase, 120 μL • 10X PCR Buffer (without magnesium), 3 x 1.25 mL • 50 mM MgCl2, 1 mL • KB Extender, 1.3 mL Store at -10°C to -30°C in a non-frost-free freezer. |
|
Low | |
Fast or Standard | |
DNA Polymerase | |
Wet or Dry Ice | |
1X | |
600 Reactions | |
Separate Components | |
DNA |
Tube | |
Platinum Taq DNA Polymerase | |
5' - 3' | |
600 reactions | |
Hot-start PCR | |
Built-In Hot Start | |
3'-A | |
5 kb or less |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.