missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Platinum™ Taq DNA Polymerase, High Fidelity

Platinum Taq DNA Polymerase, High Fidelity, is ideal for amplification of DNA fragments when high yields and robust amplification are required.

409.00 € - 9480.00 €

Specifications

Detection Method Primer-probe
GC-Rich PCR Performance Low
Polymerase Platinum Taq DNA Polymerase High Fidelity
Reaction Speed Standard
Shipping Condition Dry Ice
View More Specs

Includes: Platinum™ Taq DNA Polymerase High Fidelity (20μL, at 5U/μL) is supplied with a vial of 10X High Fidelity Buffer [600mM Tris-SO4 (pH 8.9), 180mM (NH4)2SO4] (1.25mL), and a vial of 50mM MgSO4 (1mL).

Products 3
Product Code Brand No. of Reactions Price Quantity & Availability  
Product Code Brand No. of Reactions Price Quantity & Availability  
10524253
View Documents
Invitrogen™
11304011
100 Reactions
409.00 €
100 reactions
Please to purchase this item. Need a web account? Register with us today!
10665783
View Documents
Invitrogen™
11304029
500 Reactions
1740.00 €
500 reactions
Please to purchase this item. Need a web account? Register with us today!
10675783
View Documents
Invitrogen™
11304102
5000 Reactions
9480.00 €
5000 reactions
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Platinum Taq DNA Polymerase, High Fidelity, is ideal for amplification of DNA fragments when high yields and robust amplification are required. High fidelity is provided by a mixture of Platinum Taq DNA Polymerase and the proofreading enzyme. PCR specificity is improved with built-in Platinum hot-start technology.

Features of Platinum Taq DNA Polymerase, High Fidelity

  • Greater than six times higher fidelity than Taq DNA polymerase
  • Amplification of fragments up to 15 kb
  • Convenient room temperature reaction assembly
  • Increased specificity, and yield

Applications

  • Amplification of complex DNA templates
  • Amplification of viral and plasmid templates
  • RT-PCR

Order Info

Shipping Condition: Dry ice

Specifications

Specifications

Primer-probe
Platinum Taq DNA Polymerase High Fidelity
Dry Ice
6X
Separate Components
Low
Standard
Hot-start PCR, High-fidelity PCR
Built-In Hot Start
20 kb or less
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only. Not for use in diagnostic procedures.