Learn More
SuperScript™, Platinum™ SuperScript™ III Platinum™ One-Step qRT-PCR Kit w/ROX
Description
Includes
SuperScript™ III/Platinum™ Taq Mix, 2X Reaction Mix with ROX and 1mL Magnesium Sulfate (50mM). Guaranteed stable for 6 months when properly stored.
One-Step qRT-PCR Kits with ROX combine the most powerful reverse transcriptase and DNA polymerase technologies to deliver precise and accurate analysis of gene expression in a convenient, high-throughput one-step format. One-Step qRT-PCR kits with ROX include SuperScript™ III Reverse Transcriptase which produces higher yields of full-length cDNA for broader gene representation, Platinum™ Taq DNA Polymerase which activates faster and more completely than other hot-start enzymes to deliver more sensitive amplification and ROX reference dye premixed at optimal concentration for ABI instruments
- SuperScript™ III Platinum™ SYBR™ Green One-Step qRT-PCR Kit provides a highly efficient, cost-effective qRT-PCR system with SYBR Green I detection
- SuperScript™ III Platinum™ One-Step qRT-PCR Kit provides sensitive, specific detection for use with LUX™ fluorogenic primers or dual-labeled probes (e.g. TaqMan™)
Order Info
Dry Ice
Specifications
Specifications
| Content And Storage | • 500 μL SuperScript™ III/Platinum™ Taq Mix • 12.5 mL 2X Reaction Mix with ROX • 2 × 1 mL Magnesium Sulfate (50 mM) Store components in the dark at -20°C. |
| Detection Method | Primer-probe |
| PCR Method | 1-step RT-qPCR |
| Polymerase | Platinum Taq DNA Polymerase |
| Reaction Speed | Standard |
| For Use With (Equipment) | 7000 System, 7300 System, 7700 System, 7900HT System |
| Product Line | Platinum, SuperScript |
| Product Type | One-Step qRT-PCR Kit |
| Quantity | 500 reactions |
| Shipping Condition | Dry Ice |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.