missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SuperScript™ III One-Step RT-PCR System with Platinum™ Taq DNA Polymerase

The SuperScript III One-Step RT-PCR System with Platinum Taq DNA Polymerase is designed for the sensitive, reproducible, end-point detection and analysis of RNA molecules by RT-PCR.

352.00 € - 1046.00 €

Specifications

Detection Method Gel Electrophoresis
Format One-Step RT-PCR System
GC-Rich PCR Performance High
PCR Method 1-step RT-PCR
Polymerase Platinum Taq DNA Polymerase
View More Specs

Products 2
Product Code Brand No. of Reactions Price Quantity & Availability  
Product Code Brand No. of Reactions Price Quantity & Availability  
10464022
View Documents
Invitrogen™
12574018
25 Reactions
352.00 €
25 reactions
Please to purchase this item. Need a web account? Register with us today!
10082922
View Documents
Invitrogen™
12574026
100 Reactions
1046.00 €
100 reactions
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The SuperScript III One-Step RT-PCR System with Platinum Taq DNA Polymerase is designed for the sensitive, reproducible, end-point detection and analysis of RNA molecules by RT-PCR. Using this convenient one-step formulation, you can perform both cDNA synthesis and PCR amplification in a single tube using gene-specific primers and target RNAs from either total RNA or mRNA. The system uses a mixture of SuperScript III Reverse Transcriptase and Platinum Taq DNA polymerase in an optimized reaction buffer, and it can detect a wide range of RNA targets, from 200 bp to 4.5 kb. The amount of starting material can range from 0.01 pg to 1 μg of total RNA.

Key advantages of SuperScript III One-Step RT-PCR System with Platinum Taq DNA Polymerase
• Sensitive detection down to 0.01 pg total RNA
• Convenient one-step format for increased speed, convenience, and less reaction-to-reaction variability
• SuperScript III RT for cDNA synthesis up to 55°C, for more specific priming with gene specific primers
• Amplicon size—compatible detection of targets up to 4.5 kb in length for greater flexibility

SuperScript III Reverse Transcriptase
SuperScript III Reverse Transcriptase is a version of M-MLV RT that has been engineered to reduce RNase H activity and provide increased thermal stability. The enzyme can synthesize cDNA at a temperature range of 45–60°C, providing increased specificity, higher yields of cDNA, and more full-length product than other reverse transcriptases. Because SuperScript III RT is not significantly inhibited by ribosomal and transfer RNA, it can be used to synthesize cDNA from total RNA.

Platinum Taq DNA Polymerase
Platinum Taq DNA Polymerase is recombinant Taq DNA polymerase complexed with a proprietary antibody that blocks polymerase activity at ambient temperatures. Activity is restored after the denaturation step in PCR cycling at 94°C, providing an automatic 'hot start' in PCR for increased sensitivity, specificity, and yield.

Note: This kit has been optimized for end-point RT-PCR. For RT–qPCR in this product line, use the SuperScript III Platinum One–Step Quantitative RT-PCR System.

Order Info

Shipping Condition: Dry ice

Specifications

Specifications

Gel Electrophoresis
High
Platinum Taq DNA Polymerase
1-Step RT-PCR
SuperScript III
Dry Ice
Built-In Hot Start
Reverse Transcription
RNA
One-Step RT-PCR System
1-step RT-PCR
30 min.
50°C
Reduced
PCR Amplified cDNA
Premixed Components
Up to 4.5 kb
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only. Not for use in diagnostic procedures.