missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ pPIC9K Pichia Vector
Carries the kanamycin resistance gene that confers resistance to Geneticin Reagent in Pichia
Brand: Invitrogen™ V17520
851.76 EUR valid until 2024-12-31
Use promo code "21651" to get your promotional price.
This item is not returnable.
View return policy
Description
- Spontaneous generation of multiple insertion events can be identified by resistance to increased levels of Geneticin Reagent
- Pichia transformants are selected on histidine-deficient medium and screened for their level of resistance to Geneticin Reagent
- Ability to grow in high concentrations of Geneticin indicates that multiple copies of kanamycin resistance gene and gene of interest are integrated into genome
- pPIC9K vector directs secretion of expressed proteins
Pichia pastoris Expression Systems, Protein Expression, Proteins, Expression, Isolation and Analysis, Yeast Expression
Specifications
AOX1 | |
Store at -20°C. | |
Untagged | |
20 μg |
Pichia Expression Vector | |
Ampicillin (AmpR), Gentamicin (GmR) | |
Restriction Enzyme/MCS | |
pPIC |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?Share a Content Correction