Learn More
Invitrogen™ pPICZα A, B, & C Pichia Vectors
Allows high-level, methanol inducible expression of the gene of interest in Pichia, and can be used in Pichia strains including X-33, SMD1168H, and KM71H
Brand: Invitrogen™ V19520
974.10 EUR valid until 2024-09-27
Use promo code "21650" to get your promotional price.
Includes: 20μg each of lyophilized pPICZ A, B, and C is supplied
Description
- Recombinant proteins are expressed as fusions to an N-terminal peptide encoding Saccharomyces cerevisiae '-factor secretion signal
- pPICα vectors contain AOX1 promoter
- All three reading frames (A, B, C versions) are provided to facilitate in-frame cloning with C-terminal peptide
- α-factor secretion signal for directing secreted expression of recombinant protein
- Zeocin resistance gene for selection in both E. coli and Pichia
- C-terminal peptide containing c-myc epitope and polyhistidine (6xHis) tag for detection and purification of recombinant fusion protein
Pichia pastoris Expression Systems, Protein Expression, Proteins, Expression, Isolation and Analysis, Yeast Expression
Specifications
AOX1 | |
20 μg each of lyophilized pPICZ A, B, and C is supplied (Store at -20C) | |
Room Temperature | |
Restriction Enzyme/MCS | |
pPIC |
Pichia Expression Vector | |
Zeocin™ (ZeoR) | |
His Tag (6x), c-Myc Epitope Tag | |
20 μg |
For Research Use Only. Not for use in diagnostic procedures.