missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human IL-6 ProQuantum Immunoassay Kit
GREENER_CHOICE

ELISA

788.00 € - 3530.00 €

Specifications

Accession Number P05231
Assay Range 0.064 to 10000 pg/mL
Assay Sensitivity <0.05 pg/mL
Conjugate FAM™
Detection Method Taqman™ Real-time PCR
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15884442
GREENER_CHOICE
View Documents
Invitrogen™
A35573
96 Tests
788.00 €
Each
Estimated Shipment: 14-04-2025
to see stock.
Add to basket
 
15926715
GREENER_CHOICE
View Documents
Invitrogen™
A355735
5 x 96 Tests
3530.00 €
Pack of 5
Estimated Shipment: 11-04-2025
to see stock.
Add to basket
 
Description

Description

IL-6 is a gene that encodes a cytokine that plays a role in inflammation and the maturation of B cells. The protein produced by this gene is an endogenous pyrogen that can induce fever in people with autoimmune diseases or infections. The gene is primarily expressed at sites of acute and chronic inflammation, where it is secreted into the serum and induces a transcriptional inflammatory response through interleukin 6 receptor, alpha. The functioning of this gene is implicated in a wide variety of inflammation-associated disease states, including suspectibility to diabetes mellitus and systemic juvenile rheumatoid arthritis. Elevated levels of the encoded protein have been found in virus infections, including COVID-19 (disease caused by SARS-CoV-2). Diseases associated with IL6 include Kaposi Sarcoma and Rheumatoid Arthritis, Systemic Juvenile.
Specifications

Specifications

P05231
<0.05 pg/mL
Taqman™ Real-time PCR
Plasma, Serum, Supernatant
Cytokine Protein Quantitation
3569
Assay Dilution Buffer, Antibody-Conjugate A, Antibody-Conjugate B, Antibody-Conjugate Dilution Buffer, Standard, Master Mix, Ligase
RUO
-20°C
30 mins
0.064 to 10000 pg/mL
FAM™
ProQuantum
Human
qPCR Instruments
BSF-2; BSF2; CDF; HGF; HSF; IFN-beta-2; IFNB2; IL-6
FAM™
Plasma 2-5 μL, Serum 2-5 μL, Supernatant 2-5 μL
IL-6
2 hrs
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.