missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IL-8 Human ProQuantum Immunoassay Kit
Measure cytokine proteins with small sample consumption and high sensitvity on a qPCR instrument
Brand: Invitrogen™ A35575
678.30 EUR valid until 2024-12-31
Use promo code "21989" to get your promotional price.
This item is not returnable.
View return policy
Description
IL-8 (Interleukin-8) is a 72 amino acid pro-inflammatory CXC chemokine designated CXCL8,secreted by endothelial cells and monocytes,which acts as a major chemoattractant for neutrophils and also T cells and basophils. IL-8,first isolated from osteosarcoma cells,contains the ELR-motif (N-terminal Glu-Leu-Arg amino acid sequence) and signals through the CXCR1 and CXCR2 receptors. Cancer studies have demonstrated a role for IL-8 in the angiogenesis and growth of tumours. Previous nomenclature for IL-8 includes neutrophil activating protein 1 (NAP-1),granulocyte chemotactic protein 1 (GCP-1),monocyte-derived neutrophil-activating peptide (MONAP) and protein 3-10C.Specifications
P10145 | |
<0.01pg/mL | |
ProQuantum | |
Human | |
qPCR Instruments | |
GCP-1; GCP1; IL8; LECT; LUCT; LYNAP; MDNCF; MONAP; NAF; NAP-1; NAP1 | |
FAM™ | |
RUO | |
-20°C | |
30 min. |
0.0128 to 5000 pg/mL | |
Taqman™ Real-time PCR | |
Plasma, Serum, Supernatant | |
Cytokine protein quantitation | |
3576 | |
Assay Dilution Buffer<Br>Antibody-Conjugate A<Br>Antibody-Conjugate B<Br>Antibody-Conjugate Dilution Buffer<Br>Standard<Br>Master Mix<Br>Ligase | |
96 Tests | |
Serum, 2-5 μL; Plasma, 2-5 μL; Supernatant, 2-5 μL | |
Human | |
2 hr. |