missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ IL-8 Human ProQuantum Immunoassay Kit
GREENER_CHOICE

Measure cytokine proteins with small sample consumption and high sensitvity on a qPCR instrument

Brand:  Invitrogen™ A355755

3044.75 EUR valid until 2024-12-31
Use promo code "21989" to get your promotional price.


 View more versions of this product

Product Code. 17133099

  • 3350.00 € / Each
Estimated Shipment: 30-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

IL-8 (Interleukin-8) is a 72 amino acid pro-inflammatory CXC chemokine designated CXCL8,secreted by endothelial cells and monocytes,which acts as a major chemoattractant for neutrophils and also T cells and basophils. IL-8,first isolated from osteosarcoma cells,contains the ELR-motif (N-terminal Glu-Leu-Arg amino acid sequence) and signals through the CXCR1 and CXCR2 receptors. Cancer studies have demonstrated a role for IL-8 in the angiogenesis and growth of tumours. Previous nomenclature for IL-8 includes neutrophil activating protein 1 (NAP-1),granulocyte chemotactic protein 1 (GCP-1),monocyte-derived neutrophil-activating peptide (MONAP) and protein 3-10C.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

P10145
<0.01pg/mL
ProQuantum
Human
qPCR Instruments
GCP-1; GCP1; IL8; LECT; LUCT; LYNAP; MDNCF; MONAP; NAF; NAP-1; NAP1
FAM™
RUO
-20°C
30 min.
0.0128 to 5000 pg/mL
Taqman™ Real-time PCR
Plasma, Serum, Supernatant
Cytokine protein quantitation
3576
Assay Dilution Buffer<Br>Antibody-Conjugate A<Br>Antibody-Conjugate B<Br>Antibody-Conjugate Dilution Buffer<Br>Standard<Br>Master Mix<Br>Ligase
5 x 96 Tests
Serum, 2-5 μL; Plasma, 2-5 μL; Supernatant, 2-5 μL
Human
2 hr.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers