missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PureLink™ Genomic DNA Mini Kit

The PureLink™ Genomic DNA Mini Kit enables high-yield, high-purity, genomic DNA (gDNA) extractions from a wide variety of sample types. Use this single kit for genomic DNA purification from blood, tissues, cells, bacteria, swabs, and blood spots, with a familiar silica-based, microcentrifuge spin-column format.

Brand:  Invitrogen™ K182002

678.00 EUR valid until 2024-12-31
Use promo code "21651" to get your promotional price.



 View more versions of this product

Product Code. 10458843

  • 904.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

The PureLink™ Genomic DNA Mini Kit enables high-yield, high-purity, genomic DNA (gDNA) extractions from a wide variety of sample types. Use this single kit for genomic DNA purification from blood, tissues, cells, bacteria, swabs, and blood spots, with a familiar silica-based, microcentrifuge spin-column format. Features of the PureLink™ Genomic DNA Mini Kit include:

Kit flexibility—one kit works with a variety of sample types and sizes
Ultraclean gDNA—minimal contamination of DNA product means successful downstream application
Improved design—optimized spin-column design and buffer formulation for better yield and purity

Multiple sample sources, one kit
The PureLink™ Genomic DNA Mini Kit can be used with many different sample types, each with its own optimized protocol outlined in the manual.

Optimized components and protocols
The PureLink™ Genomic DNA Mini Kit includes parts and protocols that improve upon the easy and familiar spin-column methods currently in use. Columns are newly configured to bind and release DNA more efficiently. Each sample source uses a specialized lysis procedure and an optimized lysis buffer formulated to enhance proteinase K activity and eliminate protein contamination. A chaotropic salt buffer promotes stable binding of DNA to the column resin, and powerful wash buffers remove all traces of protein and salt. DNA is eluted in a low-salt buffer to allow for pH stabilization of the DNA in storage.

Higher yields and purity
With PureLink™ Genomic DNA Kits you can expect high yields of high-purity gDNA (determined by A260/A280 measurements), no matter the sample type, from bacteria to tissues to blood to cultured cells. The enhanced PureLink™ Lysis and Wash Buffers help to consistently deliver purified gDNA samples with absorbance reading ratios of ∼1.9 for both A260/A280 and A260/A230, indicating no detectable contamination from carryover of protein or guanidine.

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY
Specifications

Specifications

• 50 mL PureLink Genomic Lysis/Binding Buffer; room temperature
• 45 mL PureLink Genomic Digestion Buffer; room temperature
• 50 mL PureLinkGenomic Wash Buffer 1; room temperature
• 37.5 mL PureLink Genomic Wash Buffer 2; room temperature
• 50 mL PureLink Genomic Elution Buffer; room temperature
• 5 mL RNase A (20 mg/mL); room temperature
• 5 mL Proteinase K (20 mg/mL); room temperature, or 4°C for long-term storage
• 5 x 50 PureLink Spin Columns with Collection Tubes; room temperature
• 5 x 100 PureLink Collection Tubes; room temperature

Mini
25 to 200 μL
PCR, Southern blotting, sequencing, nucleic acid labeling, hybridization
250 Preps
Up to 15 min. after cell lysis
Bacteria: ≤109 cells
Cells: ≤106
Tissue: ≤25 mg
Mouse Tail: 1 cm
Rat Tail: 0.5 cm
Whole Blood: ≤1 mL
Yeast: ≤107 cells
FFPE: ≤8 sections of 5-15 μM thickness
Oragene™ Preserved Saliva: ≤4 mL
Blood Spots: ≤5 punches
Silica Spin Column
Bacteria, Cells, Dried Blood Spots, FFPE (Formalin-fixed paraffin-embedded), Mouse/Rat Tail, Saliva, Buccal Swab, Tissue, Whole Blood, Yeast
gDNA
Not High-throughput Compatible (Manual)
Room Temperature
≤40 μg
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • PURELINK GENOMIC DNA KIT (250)

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1

Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash hands thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P272-Contaminated work clothing should not be allowed out of the workplace.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P330-Rinse mouth.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : Guanidine hydrochloride, Proteinase, tritirachium album serine

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.