missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PureLink™ Nucleic Acid Purification Rack

Product Code. 10175312
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10175312

Brand: Invitrogen™ K210013

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes

The PureLink Nucleic Acid Purification Rack consists of a: Column holder rack (for processing 12 miniprep, 8 midiprep, and 4 maniprep columns), Collection Tube Rack (capable of accommodating various types and sizes of recovery tubes), and Large capacity Waste Tray for collecting waste.

Waste reservoir nests into collection tube rack for compact storage.

  • Convenient large capacity fits multiple-sized column
  • Compact nesting feature is convenient for storage
  • PureLink Nucleic Acid Purification Rack is designed for use with columns in the PureLink HiPure Plasmid Purification Systems
  • Rack includes both Collection Tube Rack for holding various sized recovery tubes and a large-capacity Waste Reservoir
  • One rack holds as many as 12 miniprep columns, 8 midiprep columns, and 4 maxiprep columns

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Product Type Nucleic Acid Purification Rack
Format Rack
Content And Storage The PureLink™ Nucleic Acid Purification Rack includes:
• Column Holder Rack (for processing 12 miniprep, 8 midiprep, and 4 maxiprep columns)
• Collection Tube Rack (capable of accommodating various types and sizes of recovery tubes)
• Large Capacity Waste Tray (for collecting waste)
Product Line PureLink
Quantity 1 Ea.
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.