missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PureLink™ Quick Gel Extraction Kit
GREENER_CHOICE

The PureLink™ Quick Gel Extraction Kit allows you to rapidly and efficiently purify DNA fragments from TAE or TBE agarose gels of various percentages.

Brand:  Invitrogen™ K210025

 View more versions of this product

Product Code. 10114993

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers

Includes: Gel Solubilization Buffer (L3)
Wash Buffer (W1)
Elution Buffer (E5); (10 mM Tris-HCl, pH 8.5)
Quick Gel Extraction Columns
Wash Tubes
Recovery Tubes
This item is not returnable. View return policy

Description

Description

The PureLink™ Quick Gel Extraction Kit allows you to rapidly and efficiently purify DNA fragments from TAE or TBE agarose gels of various percentages. DNA can be extracted and purified from agarose gels with different melting points in ∼30 minutes using PureLink™ silica membrane-based quick gel extraction columns. For your convenience, purification protocols are provided for centrifugation and for vacuum.

Advantages of using PureLink™ Quick Gel Extraction Kit:
• Purify DNA fragments from TAE and TBE agarose gels of various percentages and melting points
• Complete the procedure in ∼30 minutes
• Easily purify DNA fragments from 40 bp to 10 kb from gels
• Obtain high recovery of DNA fragments
• Bind and purify up to 15 μg DNA with one column
• Purify DNA fragments that are high quality and show reliable performance in PCR, restriction enzyme digestion, cloning, and labeling

Note: The PureLink™ Quick Gel Extraction Kit is not designed to purify supercoiled plasmid DNA or genomic DNA from agarose gels. Only linear DNA fragments may be purified from gels using this kit.

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY
Specifications

Specifications

All components of the PureLink™ Quick Gel Extraction System are shipped at room temperature. Upon receipt, store all components at room temperature.

Components:
• Gel Solubilization Buffer (L3)
• Wash Buffer (W1)
• Elution Buffer (E5); (10 mM Tris-HCl, pH 8.5)
• Quick Gel Extraction Columns
• Wash Tubes
• Recovery Tubes

Spin or Vacuum Column, Silica Spin Column
Next-Generation Sequencing, Cloning, Sequencing, Nucleic Acid Labeling, PCR, In Vitro Transcription
Not High-throughput Compatible (Manual)
PureLink™
250 Preps
Nucleic Acids in Gel Fragments
Kit
DNA Fragments in Agarose Gel Slices, Gel Samples
Less hazardous, sustainable packaging
Ethidium Bromide, SYBR Safe
Quick Gel Extraction Kit
Room Temperature
30 min.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.