Learn More
Invitrogen™ PureLink™ Quick Gel Extraction Kit
The PureLink™ Quick Gel Extraction Kit allows you to rapidly and efficiently purify DNA fragments from TAE or TBE agarose gels of various percentages.
Brand: Invitrogen™ K210012
Alert:
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21626” to get your promotional price
Includes: Gel Solubilization Buffer (L3)
Wash Buffer (W1)
Elution Buffer (E5); (10 mM Tris-HCl, pH 8.5)
Quick Gel Extraction Columns
Wash Tubes
Recovery Tubes
Description
The PureLink™ Quick Gel Extraction Kit allows you to rapidly and efficiently purify DNA fragments from TAE or TBE agarose gels of various percentages. DNA can be extracted and purified from agarose gels with different melting points in ∼30 minutes using PureLink™ silica membrane-based quick gel extraction columns. For your convenience, purification protocols are provided for centrifugation and for vacuum.
Advantages of using PureLink™ Quick Gel Extraction Kit:
• Purify DNA fragments from TAE and TBE agarose gels of various percentages and melting points
• Complete the procedure in ∼30 minutes
• Easily purify DNA fragments from 40 bp to 10 kb from gels
• Obtain high recovery of DNA fragments
• Bind and purify up to 15 μg DNA with one column
• Purify DNA fragments that are high quality and show reliable performance in PCR, restriction enzyme digestion, cloning, and labeling
Note: The PureLink™ Quick Gel Extraction Kit is not designed to purify supercoiled plasmid DNA or genomic DNA from agarose gels. Only linear DNA fragments may be purified from gels using this kit.
Order Info
Shipping Condition: Room Temperature
Specifications
• 2 x 90 mL Gel Solubilization Buffer L3; room temperature • 16 mL Wash Buffer W1; room temperature • 15 mL Elution Buffer E5; room temperature • 50 Quick Gel Extraction Columns with Collection Tubes; room temperature • 50 Recovery Tubes; room temperature |
|
Agarose Gels, Gel Samples | |
DNA | |
Less hazardous, sustainable packaging | |
Ethidium Bromide, SYBR Safe | |
Room Temperature | |
15 μg (Binding capacity) |
Silica Spin Column | |
50 μL | |
PCR, Southern blotting, sequencing, nucleic acid labeling, hybridization | |
Not High-throughput Compatible (Manual) | |
50 Preps | |
30 min. | |
≤400 mg |
For Research Use Only. Not for use in diagnostic procedures.