missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Qubit™ ssDNA Assay Kit

Quantitate oligonucleotides or long ssDNA easily and quickly with the Qubit ssDNA Assay Kit, which works with the Qubit Fluorometer and other fluorometers or plate readers. Common contaminants such as salts, solvents, detergents, and protein are well tolerated with this assay.

Brand:  Invitrogen™ Q10212

138.70 EUR valid until 2025-12-31
Use promo code "24344" to get your promotional price.


Product Code. 10387943

  • 159.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

Requires : Qubit 1.0 or 2.0 Fluorometer or other fluorometer and 500μL thin-walled PCR tubes
This item is not returnable. View return policy

Description

Description

The Qubit ssDNA Assay Kit is designed specifically for use with the Qubit Fluorometer, but may be used with any fluorometer or fluorescence plate reader. The Qubit™ ssDNA Kit is ideal for quantitating single-stranded DNA or oligonucleotides. However, it is not specific for single-stranded DNA. This assay kit will also detect double-stranded DNA and RNA but it will not detect contaminating protein or nucleotides. The assay is designed to be accurate for initial sample concentrations from 50 pg/μL to 200 ng/μL providing an assay range of 1–200 ng. The kit provides concentrated assay reagent, dilution buffer, and pre-diluted DNA standards. Simply dilute the reagent using the buffer provided, add your sample (any volume between 1 μl and 20 μl is acceptable), and read the concentration using the Qubit Fluorometer. Common contaminants, such as salts, free nucleotides, solvents, detergents, or protein are well tolerated in the assay.

Which product to choose for ssDNA quantitation?
  • For 1–20 samples: use this Qubit ssDNA Assay Kit with the Qubit Fluorometer
  • For 20–2000 samples: use the Quant-iT OliGreen ssDNA Assay Kit with microplate reader
Notes:
1. All Qubit assay kits can be used with the Qubit 1.0, Qubit 2.0, Qubit 3, and Qubit 4 fluorometers.
2. 500 μL thin-walled PCR tubes are required but not included.

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY
Specifications

Specifications

ssDNA Quantitation
500/525
0.2 to 240 ng
1 kit
Fluorescence
Qubit Fluorometer
Qubit
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number3071386 Certificate TypeCertificate of Analysis Date20/11/2024 Catalog NumberQ10212,Q10212_LITL18 PROMO
Lot Number3011754 Certificate TypeCertificate of Analysis Date22/10/2024 Catalog NumberQ10212,Q10212_LITL18 PROMO
Lot Number2980681 Certificate TypeCertificate of Analysis Date12/09/2024 Catalog NumberQ10212,Q10212_LITL18 PROMO
Lot Number2942292 Certificate TypeCertificate of Analysis Date22/08/2024 Catalog NumberQ10212,Q10212_LITL18 PROMO
Lot Number2920612 Certificate TypeCertificate of Analysis Date09/08/2024 Catalog NumberQ10212,Q10212_LITL18 PROMO
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ Qubit™ ssDNA Assay Kit > 1 kit

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.