missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Select Agar™, powder

Agar is a mixture of polysaccharides extracted from red seaweed cell walls. It has unique gelling properties that are utilized in many industrial and laboratory applications.

Brand:  Invitrogen™ 30391023

171.76 EUR valid until 2024-12-20
Use promo code "21651" to get your promotional price.


 View more versions of this product

Product Code. 11508926

  • 226.00 € / 500g
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Agar is a mixture of polysaccharides extracted from red seaweed cell walls. It has unique gelling properties that are utilized in many industrial and laboratory applications. Solid culture medium is created by adding  agar to media broth (typical final concentration is 1–2%). Solid media cell culture allows the isolation of individual E. coli colonies and is fundamental in cloning workflows.

Agar is used in diagnostic laboratories to detect pathogens from contaminated food or water. Also, it can be used to prepare selective media for saccharomyces cerevisiae and other yeasts used in various molecular microbiology procedures (e.g., YPD Agar). In addition, agar has been used in tissue double embedding for histopathology, hydrogel creation, and tissue engineering applications.

Primary applications
• Making solid media to grow cells (e.g., bacteria, fungi, yeast)
• To isolate E. coli clones with plasmid on a selective media
• Tissue engineering, hydrogel production

Look for more media that fit your needs
We offer other ready-to-use media formulations, such as LB (Lennox) (Cat. No. 10855001), Terrific Broth (Cat. No. A1374301) or S.O.C recovery medium (Cat. No. 15544034).

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Escherichia coli, Saccharomyces cerevisiae
Bottle
Autoclave
Coliforms, Fungi
Room Temperature
• 1 x 500 g bottle Select Agar Powder

Store at room temperature.

Select Agar
Dehydrated Culture Media
500 g
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.