Learn More
Invitrogen™ SequenceRx Enhancer Solution A
Designed to improve length and quality of sequencing data from difficult sequencing templates
Brand: Invitrogen™ 12238010
329.84 EUR valid until 2025-04-18
Use promo code "24107" to get your promotional price.
Description
The SequenceRx Enhancer System is a panel of seven co-solvents which are added individually to automated fluorescent DNA sequencing reactions. This system is not designed for use on long stretches of poly A.
- Useful on templates containing regions with secondary structure such as high GC-content, small nucleotide repeats, hairpins, direct repeats, and inverted repeats
- Requires no change in protocols-the SequenceRx Solution replaces part of water added to reaction mixture
- Compatible with all sequencing chemistries and platforms
- Uses one reaction to determine sequence for all four nucleotides (100 reactions equal 100 templates)
Sanger Sequencing, Sanger Sequencing Technology and Accessories, Sequencing
Order Info
Approved for shipment on Wet or Dry ice

Specifications
Store at -20°C. Guaranteed stable for 6 months when properly stored. | |
Sequencing | |
Enhancer Solution | |
Approved for shipment on Wet or Dry Ice |
Sequencing Instruments (Capillary Electrophoresis-Based) | |
200 | |
200 Reactions |
Product Suggestions
Customers who viewed this item also viewed
Safety and Handling
- SequenceR Enhancer Solution A
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : 4-methyl-morpholine-4-oxide
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
3012799 | Certificate of Analysis | 31/01/2025 | 12238010 |
2475869 | Certificate of Analysis | 29/01/2025 | 12238010 |
2509814 | Certificate of Analysis | 29/01/2025 | 12238010 |
2572437 | Certificate of Analysis | 29/01/2025 | 12238010 |
2572437 | Certificate of Analysis | 29/01/2025 | 12238010 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.